Skip to Content
Merck
All Photos(1)

Key Documents

SML1211

Sigma-Aldrich

SMANT hydrochloride

≥98% (HPLC)

Synonym(s):

N-(4-Bromophenyl)-3-(3,5-dimethylpiperidin-1-yl)propanamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23BrN2O · HCl
CAS Number:
Molecular Weight:
375.73
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

BrC1=CC=C(NC(CCN2CC(C)CC(C)C2)=O)C=C1.Cl

InChI

1S/C16H23BrN2O.ClH/c1-12-9-13(2)11-19(10-12)8-7-16(20)18-15-5-3-14(17)4-6-15;/h3-6,12-13H,7-11H2,1-2H3,(H,18,20);1H

InChI key

XQESCHFXROUCOQ-UHFFFAOYSA-N

Biochem/physiol Actions

SMANT is a Hedgehog pathway antagonist, a Smoothened antagonist that behaves similarly against normal cancer smoothened and the M2 mutant. In a study comparing activities among the currently known smoothened/Hedgehog antagonists, SMANT behaves differently, binding to another Smo site different to that bound by other compounds.
SMANT is a Hedgehog pathway antagonist; a Smoothened antagonist.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service