Skip to Content
Merck
All Photos(1)

Key Documents

SML1171

Sigma-Aldrich

GSA-10

≥98% (HPLC)

Synonym(s):

4-[[(1-Hexyl-1,2-dihydro-4-hydroxy-2-oxo-3-quinolinyl)carbonyl]amino]-benzoic acid propyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$147.00

$147.00


Estimated to ship onApril 22, 2025



Select a Size

Change View
5 MG
$147.00

About This Item

Empirical Formula (Hill Notation):
C26H30N2O5
CAS Number:
Molecular Weight:
450.53
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$147.00


Estimated to ship onApril 22, 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 0.5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

OC(C1=CC=CC=C1N2CCCCCC)=C(C(NC3=CC=C(C(OCCC)=O)C=C3)=O)C2=O

InChI

1S/C26H30N2O5/c1-3-5-6-9-16-28-21-11-8-7-10-20(21)23(29)22(25(28)31)24(30)27-19-14-12-18(13-15-19)26(32)33-17-4-2/h7-8,10-15,29H,3-6,9,16-17H2,1-2H3,(H,27,30)

InChI key

MDLUYYGRCGDKGL-UHFFFAOYSA-N

Biochem/physiol Actions

GSA-10 is a novel, potent activator of the smoothened receptor (Smo).
GSA-10 is a novel, potent activator of the smoothened receptor (Smo). GSA-10 binds to Smo at a unique site compared to agonists such as SAG and purmorphamine, and is not affected by the Smo antagonist cyclopamine. GSA-10 promotes the differentiation of mesenchymal pluripotent C3H10T1/2 cells into osteoblasts.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service