Skip to Content
Merck
All Photos(1)

Key Documents

SML1148

Sigma-Aldrich

YIL-781 hydrochloride

≥98% (HPLC)

Synonym(s):

6-(4-Fluorophenoxy)-2-methyl-3-[[(3S)-1-(1-methylethyl)-3-piperidinyl]methyl]-4(3H)-quinazolinone hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$159.00
25 MG
$634.00

$159.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$159.00
25 MG
$634.00

About This Item

Empirical Formula (Hill Notation):
C24H28FN3O2 · xHCl
CAS Number:
Molecular Weight:
409.50 (free base basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$159.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

, white to yellow to light brown

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

CC(C)N(CCC1)C[C@H]1CN(C(C)=N2)C(C3=C2C=CC(OC4=CC=C(F)C=C4)=C3)=O.Cl

InChI

1S/C24H28FN3O2.ClH/c1-16(2)27-12-4-5-18(14-27)15-28-17(3)26-23-11-10-21(13-22(23)24(28)29)30-20-8-6-19(25)7-9-20;/h6-11,13,16,18H,4-5,12,14-15H2,1-3H3;1H/t18-;/m0./s1

InChI key

QQGRMONBDBMLSP-FERBBOLQSA-N

Application

YIL-781 has been used as a ghrelin receptor (GHSR) antagonist in combination with methanandamide (mAEA) to study its effect on gastric vagal afferent mechanosensitivity.[1]

Biochem/physiol Actions

YIL-781 is a Ghrelin receptor antagonist.
YIL-781 is a potent antagonist (Ki = 17 nM) of the growth hormone secretagogue receptor (GHS-R1). YIL-781 blocks ghrelin-induced reduction of insulin secretion in rat islets. In vivo, the compound YIL-781 improves glucose tolerance and reduces weight gain.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service