Skip to Content
Merck
All Photos(1)

Key Documents

SML1095

Sigma-Aldrich

PKI-166

≥98% (HPLC)

Synonym(s):

(R)-4-[4-[(1-Phenylethyl)amino]-1H-pyrrolo[2,3-d]pyrimidin-6-yl]-phenol, (R)-6-(4-Hydroxyphenyl)-4-[(1-phenylethyl)amino]-7H-pyrrolo[2,3-d]pyrimidine, 4-[4-(((R)-1-Phenylethyl)amino)-7H-pyrrolo[2,3-d]pyrimidin-6-yl]phenol, 4-[4-[[(1R)-1-Phenylethyl]amino]-1H-pyrrolo[2,3-d]pyrimidin-6-yl]-phenol (9CI), CID 6918403, PKI 166, PKI166

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$159.00

$159.00


Estimated to ship onMay 30, 2025



Select a Size

Change View
5 MG
$159.00

About This Item

Empirical Formula (Hill Notation):
C20H18N4O
CAS Number:
Molecular Weight:
330.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$159.00


Estimated to ship onMay 30, 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(C=C1)=CC=C1C2=CC3=C(N2)N=CN=C3N[C@H](C)C4=CC=CC=C4

InChI

1S/C20H18N4O/c1-13(14-5-3-2-4-6-14)23-19-17-11-18(24-20(17)22-12-21-19)15-7-9-16(25)10-8-15/h2-13,25H,1H3,(H2,21,22,23,24)/t13-/m1/s1

InChI key

XRYJULCDUUATMC-CYBMUJFWSA-N

Gene Information

human ... EGFR(1956)

Biochem/physiol Actions

PKI-166 is a potent, selective and orally available EGF-R tyrosine kinase inhibitor.
PKI-166 is a potent, selective and orally available EGF-R tyrosine kinase inhibitor. PKI-166 potently inhibits the growth and metastasis of many human cancers including human pancreatic cancer.

Other Notes

Light and air sensitive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service