Skip to Content
Merck
All Photos(1)

Key Documents

SML1070

Sigma-Aldrich

SR-58611A

≥98% (HPLC)

Synonym(s):

Amibegron, [[(7S)-7-[[(2R)-2-(3-Chlorophenyl)-2-hydroxyethyl]amino]-5,6,7,8-tetrahydro-2-naphthalenyl]oxy acetic acid ethyl ester hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$166.00
25 MG
$653.00

$166.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$166.00
25 MG
$653.00

About This Item

Empirical Formula (Hill Notation):
C22H26ClNO4 · HCl
CAS Number:
Molecular Weight:
440.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$166.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -75 to -95°, c = 0.5 in methanol

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

ClC1=CC([C@@H](O)CN[C@@H]2CC(C=C(OCC(OCC)=O)C=C3)=C3CC2)=CC=C1.Cl

InChI

1S/C22H26ClNO4.ClH/c1-2-27-22(26)14-28-20-9-7-15-6-8-19(11-17(15)12-20)24-13-21(25)16-4-3-5-18(23)10-16;/h3-5,7,9-10,12,19,21,24-25H,2,6,8,11,13-14H2,1H3;1H/t19-,21-;/m0./s1

InChI key

NQIZCDQCNYCVAS-RQBPZYBGSA-N

Gene Information

human ... ADRB3(155)

Biochem/physiol Actions

SR58611A is a potent, specific 3β adrenergic receptor agonist.
SR58611A is a potent, specific b3 adrenergic receptor agonist. The EC50 for reduction of β3-mediated spontaneous motility in rat distal colon sections is 3.5 nM. Minimal anti-depressant efficacious doses for SR58611A are 0.1-0.3 mpk in rodent models.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zili Wang et al.
Journal of shoulder and elbow surgery, 30(2), 373-386 (2020-07-01)
Rotator cuff (RC) muscle atrophy and fatty infiltration (FI) are independent factors correlated with failure of attempted tendon repair in larger RC tears. However, there is no effective treatment for RC muscle atrophy and FI at this time. The recent

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service