Skip to Content
Merck
All Photos(1)

Key Documents

SML1066

Sigma-Aldrich

CYM50358

≥98% (HPLC)

Synonym(s):

N-[4-(Aminomethyl)-2,6-dimethylphenyl]-5-(2,5-dichlorophenyl)-2-furancarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$184.00
25 MG
$726.00

$184.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$184.00
25 MG
$726.00

About This Item

Empirical Formula (Hill Notation):
C20H18Cl2N2O2
CAS Number:
Molecular Weight:
389.28
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

$184.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=C(C)C=C(CN)C=C1C)C2=CC=C(C3=CC(Cl)=CC=C3Cl)O2

InChI

1S/C20H18Cl2N2O2/c1-11-7-13(10-23)8-12(2)19(11)24-20(25)18-6-5-17(26-18)15-9-14(21)3-4-16(15)22/h3-9H,10,23H2,1-2H3,(H,24,25)

InChI key

QWJOPXDAQCDRRM-UHFFFAOYSA-N

Biochem/physiol Actions

CYM50358 is a potent, selective antagonist of the sphingosine-1-phosphate receptor 4 (S1P4).
CYM50358 is a potent, selective antagonist of the sphingosine-1-phosphate receptor 4 (S1P4). CYM50358 inhibits S1P4 with an IC50 of 25 nM.

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service