Skip to Content
Merck
All Photos(1)

Key Documents

SML1041

Sigma-Aldrich

Atreleuton

≥98% (HPLC)

Synonym(s):

(R)-N-(3-(5-(4-Fluorophenylmethyl)thien-2-yl)-1-methyl-2-propynyl)-N-hydroxyurea, A8576, ABT 761, VIA-2291

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$147.00
25 MG
$507.00

$147.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$147.00
25 MG
$507.00

About This Item

Empirical Formula (Hill Notation):
C16H15FN2O2S
CAS Number:
Molecular Weight:
318.37
UNSPSC Code:
51111800
NACRES:
NA.77

$147.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +40 to +50°, c = 1 in methanol

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)Cc2[s]c(cc2)C#C[C@H](N(O)C(=O)N)C

InChI

1S/C16H15FN2O2S/c1-11(19(21)16(18)20)2-7-14-8-9-15(22-14)10-12-3-5-13(17)6-4-12/h3-6,8-9,11,21H,10H2,1H3,(H2,18,20)/t11-/m1/s1

InChI key

MMSNEKOTSJRTRI-LLVKDONJSA-N

Biochem/physiol Actions

Atreleuton is a reversible 5-lipoxygenase inhibitor.
Atreleuton is a reversible 5-lipoxygenase inhibitor. Atreleuton exhibits potent and selective inhibition of leukotriene formation both in vitro and in vivo.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service