Skip to Content
Merck
All Photos(1)

Key Documents

SML1038

Sigma-Aldrich

MKC-733

≥98% (HPLC)

Synonym(s):

DDP733, N-(3R)-1-Azabicyclo[2.2.2]oct-3-yl-4,7-dihydro-7-oxo-thieno[3,2-b]pyridine-6-carboxamide hydrochloride, Pumosetrag

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$435.00

$435.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$435.00

About This Item

Empirical Formula (Hill Notation):
C15H17N3O2S · HCl
CAS Number:
Molecular Weight:
339.84
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$435.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -15 to -20°, c = 1 in H2O

color

white to beige

solubility

H2O: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CNC(C=CS2)=C2C1=O)N[C@@H]3C4CCN(CC4)C3.Cl

InChI

1S/C15H17N3O2S.ClH/c19-13-10(7-16-11-3-6-21-14(11)13)15(20)17-12-8-18-4-1-9(12)2-5-18;/h3,6-7,9,12H,1-2,4-5,8H2,(H,16,19)(H,17,20);1H/t12-;/m0./s1

InChI key

XGVADZZDFADEOO-YDALLXLXSA-N

Biochem/physiol Actions

MKC-733 is a 5-HT3 receptor agonist.
MKC-733 is a 5-HT3 specific agonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service