Skip to Content
Merck
All Photos(1)

Key Documents

SML1019

Sigma-Aldrich

Lofexidine hydrochloride

≥98% (HPLC)

Synonym(s):

2-[1-(2,6-Dichlorophenoxy)ethyl]-4,5-dihydro-1H-imidazole hydrochloride, Baq 168, Britlofex, Lofetensin, Loxacor, MDL 14042

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$93.00
50 MG
$363.00

$93.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$93.00
50 MG
$363.00

About This Item

Empirical Formula (Hill Notation):
C11H12Cl2N2O · HCl
CAS Number:
Molecular Weight:
295.59
UNSPSC Code:
12352200
NACRES:
NA.77

$93.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C11H12Cl2N2O.ClH/c1-7(11-14-5-6-15-11)16-10-8(12)3-2-4-9(10)13;/h2-4,7H,5-6H2,1H3,(H,14,15);1H

InChI key

DWWHMKBNNNZGHF-UHFFFAOYSA-N

Biochem/physiol Actions

Lofexidine, a derivative of clonidine, is a very potent agonist of α2 adrenoceptors. Lofexidine is used as a treatment for withdrawl from heroin and other opiates, and as a short-activg anti-hypertensive agent.
Lofexidine, a derivative of clonidine, is a very potent agonist of a2 adrenoceptors.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. NEED ADDITIONAL DATA FOR LOFEXIDINE HYDROCHLORIDE CAT# SML1019 SUCH AS NMR TESTING AND ANY ADDITIONAL TESTING OTHER THAN COA.

    1 answer
    1. The only in-house release testing criteria & specifications are the ones listed on the corresponding Certificate of Analysis.

      Additional testing criteria may be possible on a "CUSTOM" basis for a “BULK” quantity, if such CUSTOM testing/specification(s) is/are deemed feasible by our Team. To further discuss, kindly ask you to navigate to the link https://www.sigmaaldrich.com/techservice, click on "Product Technical Inquires" under the Products Section with all the required information so that a member of our team can reach out to you to assist further. Thank you.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service