Skip to Content
Merck
All Photos(1)

Key Documents

SML0984

Sigma-Aldrich

7-Oxostaurosporine

≥98% (HPLC)

Synonym(s):

9,13-Epoxy-1H,9H-diindolo[1,2,3-gh:3′,2′,1′-lm]pyrrolo[3,4-j][1,7]benzodiazonine-1,3(2H)-dione, 10,11,12,13-tetrahydro-10-methoxy-9-methyl-11-(methylamino)-, (9S,10R,11R,13R)-, Antibiotic BMY 41950, Antibiotic RK 1409

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$422.00

$422.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$422.00

About This Item

Empirical Formula (Hill Notation):
C28H24N4O4
CAS Number:
Molecular Weight:
480.51
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

$422.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

O=C(NC1=O)C2=C1C3=C4C5=C2C6=C(C=CC=C6)N5[C@H](C[C@H]7NC)O[C@]([C@@H]7OC)(C)N4C8=C3C=CC=C8

InChI

1S/C28H24N4O4/c1-28-25(35-3)15(29-2)12-18(36-28)31-16-10-6-4-8-13(16)19-21-22(27(34)30-26(21)33)20-14-9-5-7-11-17(14)32(28)24(20)23(19)31/h4-11,15,18,25,29H,12H2,1-3H3,(H,30,33,34)/t15-,18+,25-,28+/m1/s1

InChI key

POTTVLREWUNNRO-MJQUMLNUSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service