Skip to Content
Merck
All Photos(1)

Documents

SML0978

Sigma-Aldrich

ML291

≥98% (HPLC)

Synonym(s):

CID52940465, N-(4-((4-Chloropiperidin-1-yl)sulfonyl)phenyl)-5-nitrofuran-2-carboxamide, SID134228465

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16ClN3O6S
CAS Number:
Molecular Weight:
413.83
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

ML291 specifically activates the apoptotic arm of the cellular unfolded protein response ( UPR) EC50 = 762 nM) with 105-fold selectivity over activation of the adaptive UPR pathway. ML291 induces apoptosis in wild-type mouse embryonic fibroblasts (MEFs), but not in CHOP KO MEFs.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service