Skip to Content
Merck
All Photos(1)

Key Documents

SML0934

Sigma-Aldrich

Enfuvirtide acetate salt

≥95% (HPLC)

Synonym(s):

T-20 acetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$148.00

$148.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$148.00

About This Item

Empirical Formula (Hill Notation):
C204H301N51O64 · xC2H4O2
CAS Number:
Molecular Weight:
4491.88 (free base basis)
UNSPSC Code:
51111800
NACRES:
NA.77

$148.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated
protect from light

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

Amino Acid Sequence

Ac-Tyr-Thr-Ser-Leu-Ile-His-Ser-Leu- Ile-Glu-Glu-Ser-Gln-Asn-Gln-Gln-Glu- Lys-Asn-Glu-Gln-Glu-Leu-Leu-Glu- Leu-Asp-Lys-Trp-Ala-Ser-Leu-Trp-Asn-Trp-Phe-NH2 acetate

General description

Enfuvirtide acetate salt is a linear synthetic peptide made of 36 amino acids. It has an acetylated N-terminus and a carboxamide C-terminus. Enfuvirtide is catabolized with the help of proteolytic enzymes.[1]

Biochem/physiol Actions

Enfuvirtide is an HIV fusion inhibitor used to patients with multi-drug resistant HIV. Enfuvirtide binds to gp41 preventing the creation of an entry pore for the HIV-1 virus.
Enfuvirtide is an HIV fusion inhibitor.

Other Notes

Light sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Desk Encyclopedia of Microbiology (2010)
Alexander Herrmann et al.
Scientific reports, 10(1), 1326-1326 (2020-01-30)
Overcoming the global health threat of HIV infection requires continuous pipelines of novel drug candidates. We identified the γ-pyrone polyketides Aureothin/Neoaureothin as potent hits by anti-HIV screening of an extensive natural compound collection. Total synthesis of a structurally diverse group

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service