Skip to Content
Merck
All Photos(1)

Key Documents

SML0886

Sigma-Aldrich

Leukadherin-1

≥98% (HPLC)

Synonym(s):

4-[5-[[4-Oxo-3-(phenylmethyl)-2-thioxo-5-thiazolidinylidene]methyl]-2-furanyl]-benzoic acid, LA1, Leukadhedrin 1, Leukadherin 1

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$114.00
25 MG
$447.00

$114.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$114.00
25 MG
$447.00

About This Item

Empirical Formula (Hill Notation):
C22H15NO4S2
CAS Number:
Molecular Weight:
421.49
UNSPSC Code:
12352200
NACRES:
NA.77

$114.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

, yellow to orange to brown

solubility

DMSO: 3 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

S1\C(=C/c3[o]c(cc3)c4ccc(cc4)C(=O)O)\C(=O)N(C1=S)Cc2ccccc2

InChI

1S/C22H15NO4S2/c24-20-19(29-22(28)23(20)13-14-4-2-1-3-5-14)12-17-10-11-18(27-17)15-6-8-16(9-7-15)21(25)26/h1-12H,13H2,(H,25,26)/b19-12-

InChI key

AEZGRQSLKVNPCI-UNOMPAQXSA-N

Biochem/physiol Actions

Leukadherin-1 is an agonist of complement receptor-3 (CR3) and exhibits anti-inflammatory activity. It is an allosteric agonist that activates integrin by binding to a different site from the ligand binding. Leukadherin-1 plays an important role in the margination and adhesion of neutrophils to the endothelial lining of the blood vessels.[1]
Leukadherin-1 is an agonist of integrin CD11b/CD18
Leukadherin-1 is an agonist of integrin CD11b/CD18 that can increase the extent of CD11b/CD18-dependent cell adhesion of transfected cells and of primary human and mouse neutrophils. Also, Leukadherin-1 decreases leukocyte recruitment and reduced arterial narrowing after injury in rats.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catherine M Dickinson et al.
Journal of intensive care, 6, 19-19 (2018-03-24)
Multi-organ failure occurs during critical illness and is mediated in part by destructive neutrophil-to-endothelial interactions. The β2 integrin receptor, CR3 (complement receptor 3; Mac-1; CD11b/CD18), which binds endothelial intercellular adhesion molecule-1 (ICAM-1), plays a key role in promoting the adhesion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service