Skip to Content
Merck
All Photos(1)

Key Documents

SML0874

Sigma-Aldrich

GMQ hydrochloride

≥95% (HPLC)

Synonym(s):

2-Guanidine-4-methylquinazoline hydrochloride, 4-Methyl-2-guanidino quinazoline hydrochloride, N-(4-Methyl-2-quinazolinyl)-guanidine hydrochloride, NSC 403387 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$131.00

$131.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$131.00

About This Item

Empirical Formula (Hill Notation):
C10H11N5 · HCl
CAS Number:
Molecular Weight:
237.69
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$131.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

CC1=NC(NC(N)=N)=NC2=C1C=CC=C2.Cl

InChI

1S/C10H11N5.ClH/c1-6-7-4-2-3-5-8(7)14-10(13-6)15-9(11)12;/h2-5H,1H3,(H4,11,12,13,14,15);1H

InChI key

VNPZWQRWOHFAPD-UHFFFAOYSA-N

Biochem/physiol Actions

GMQ/2-Guanidine-4-methylquinazoline is a potent and selective modulator of acid-sensing ion channel (ASIC) that activates ASIC3 channels under neutral pH. GQM blocks acid-induced maximal peak current and alters ASICs pH dependence for activation and inactivation.
GMQ/2-guanidine-4-methylquinazoline is a potent and selective modulator of acid-sensing ion channel (ASIC).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kojun Tsuji et al.
American journal of physiology. Gastrointestinal and liver physiology, 319(5), G564-G572 (2020-09-04)
Capsaicin powerfully evokes the swallowing reflex and is a known therapeutic agent for improving dysphagia and preventing aspiration pneumonia. However, the role of capsaicin-sensitive nerves in the initiation of swallowing evoked by various natural stimuli remains unclear. To explore this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service