Skip to Content
Merck
All Photos(1)

Key Documents

SML0857

Sigma-Aldrich

HQL 79

≥98% (HPLC)

Synonym(s):

4-Benzhydryloxy-1-[3-(1H-tetrazol-5-yl)-propyl]-piperidine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$105.00
25 MG
$510.00

$105.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$105.00
25 MG
$510.00

About This Item

Empirical Formula (Hill Notation):
C22H27N5O
CAS Number:
Molecular Weight:
377.48
UNSPSC Code:
41106300
NACRES:
NA.77

$105.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

[nH]1nnnc1CCCN2CCC(CC2)OC(c4ccccc4)c3ccccc3

InChI

1S/C22H27N5O/c1-3-8-18(9-4-1)22(19-10-5-2-6-11-19)28-20-13-16-27(17-14-20)15-7-12-21-23-25-26-24-21/h1-6,8-11,20,22H,7,12-17H2,(H,23,24,25,26)

InChI key

TZQGXAHOROZEKN-UHFFFAOYSA-N

Biochem/physiol Actions

HQL 79 is an orally active anti-allergic agent that acts as an antagonist for histamine H1 receptors (H1R), eliciting anti-asthmatic effects. It interacts with prostaglandin (PG) D synthase (H-PGDS) in the catalytic region. It may be potential lead due to its H-PGDS inhibition to treat allergies.[1]
HQL-79 is a selective inhibtor of hematopoietic prostaglandin D (PGD) synthase, one of two synthases involved in the production of Prostaglandin D2 (PGD2) from arachidonic acid. PGD2 is a lipid signaling molecule, which activates two receptors, DP1 involved in centrally mediated processes such as sleep and pain and DP2 involved and inflammation. The two PGD synthases involved in its synthesis are lipocalin-type (L-PGDS) and hematopoietic (H-PDGS), which acts to form PGD2 in mast cells, Th2 cells, microglia, and other inflammatory cells/tissues. HQL-79 selectively inhibited the activity of recombinant H-PGDS with an IC50 of 6 μM and had almost no effect on COX-1, COX-2, m-PGES, or L-PGDS up to 300 μM. HQL-79 has anti-inflammatory activity in vitro and in vivo.
HQL-79 is a selective inhibtor of hematopoietic prostaglandin D (PGD) synthase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kosuke Aritake et al.
The Journal of biological chemistry, 281(22), 15277-15286 (2006-03-21)
We determined the crystal structure of human hematopoietic prostaglandin (PG) D synthase (H-PGDS) as the quaternary complex with glutathione (GSH), Mg2+, and an inhibitor, HQL-79, having anti-inflammatory activities in vivo, at a 1.45-A resolution. In the quaternary complex, HQL-79 was

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service