Skip to Content
Merck
All Photos(1)

Key Documents

SML0846

Sigma-Aldrich

Anagrelide hydrochloride

≥98% (HPLC)

Synonym(s):

6,7-Dichloro-1,5-dihydro-imidazo[2,1-b]quinazolin-2(3H)-one hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$126.00
50 MG
$510.00

$126.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$126.00
50 MG
$510.00

About This Item

Empirical Formula (Hill Notation):
C10H7Cl2N3O · HCl
CAS Number:
Molecular Weight:
292.55
UNSPSC Code:
41106305
NACRES:
NA.77

$126.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

InChI

1S/C10H7Cl2N3O.ClH/c11-6-1-2-7-5(9(6)12)3-15-4-8(16)14-10(15)13-7;/h1-2H,3-4H2,(H,13,14,16);1H

InChI key

TVWRQCIPWUCNMI-UHFFFAOYSA-N

General description

Download the CofA, SDS and IFU specific to your product lot and format
Fab fragments from polyclonal anti-fluorescein antibodies, conjugated to alkaline phosphatase.
After immunization with fluorescein, the sheep IgG was purified by ion exchange chromatography and the specific IgG was isolated by immunosorption. The Fab fragments obtained by papain digestion were purified by gel filtration, conjugated with alkaline phosphatase, and stabilized in 50 mM triethanolamine buffer, 3 M NaCl, 1 mM MgCl2, 0.1 mM ZnCl2, 1% bovine serum albumin (w/v), pH 7.6.
Clone: polyclonal
Ig class: sheep IgG, Fab fragments
Anagrelide belongs to the imidazo[2,1-b]quinazolin-2-one series of compounds.[1]

Biochem/physiol Actions

Anagrelide is a phosphodiesterase inhibitor with antiplatelet activity (IC50 = 36 nM for inhibition of phosphodiesterase-III). Anagrelide inhibits the maturation of megakaryocytes into platelets, reducing both megakaryocyte hyperproliferation and differentiation. As a drug, anagrelide is antithrombocythemic used for the treatment of overproduction of blood platelets.
Anagrelide is a phosphodiesterase inhibitor with antiplatelet activity.
In human and animals, anagrelide has an inhibitory effect on platelet aggregation.[1] Small doses of anagrelide can generate thrombocytopenia. Anagrelide is not advised during pregnancy. It is effectively used in the treatment of essential thrombocytosis.[2]

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphodiesterases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hematology: Diagnosis and Treatment E-Book (2013)
Thrombosis and Hemorrhage (2003)
Gabriel Sollberger et al.
eLife, 9 (2020-05-12)
Neutrophils are important innate immune cells that tackle invading pathogens with different effector mechanisms. They acquire this antimicrobial potential during their maturation in the bone marrow, where they differentiate from hematopoietic stem cells in a process called granulopoiesis. Mature neutrophils

Articles

Cyclic nucleotide phosphodiesterases (PDEs) catalyze the hydrolysis of cAMP and/or cGMP. There are 11 different mammalian PDE families.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service