Skip to Content
Merck
All Photos(1)

Key Documents

SML0828

Sigma-Aldrich

FPH2

≥98% (HPLC)

Synonym(s):

4-(3-(5-Chloro-2-methoxyphenyl)thioxureido)-1-ethyl-1H-pyrazole-3-carboxamide, 4-[[[(5-Chloro-2-methoxyphenyl)amino]thioxomethyl]amino]-1-ethyl-1H-pyrazole-3-carboxamide, BRD-9424

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$158.00
25 MG
$601.00

$158.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$158.00
25 MG
$601.00

About This Item

Empirical Formula (Hill Notation):
C14H16ClN5O2S
CAS Number:
Molecular Weight:
353.83
UNSPSC Code:
12352200
NACRES:
NA.77

$158.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

S=C(Nc2c[n](nc2C(=O)N)CC)Nc1c(ccc(c1)Cl)OC

InChI

1S/C14H16ClN5O2S/c1-3-20-7-10(12(19-20)13(16)21)18-14(23)17-9-6-8(15)4-5-11(9)22-2/h4-7H,3H2,1-2H3,(H2,16,21)(H2,17,18,23)

InChI key

PCHRYHSDDPPZBV-UHFFFAOYSA-N

Biochem/physiol Actions

FPH2 (BRD-9424) induced functional proliferation of primary human hepatocytes in vitro.
FPH2 (BRD-9424) induced functional proliferation of primary human hepatocytes in vitro. Human induced pluripotent stem (iPS) cells can be differentiated toward hepatocyte-like (iHep) cells, but replication is not great, and cells typically show an immature hepatic phenotype and have limited use as a renewable source of functional human hepatocytes. FPH2 induced hepatocyte doublings at a rate consistent with reported liver regeneration kinetics. FPH2 can be used in conjunction with FH1 (BRD-K4477), which induces a more mature phenotype.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service