Skip to Content
Merck

Skip To

SML0803

VUF10661

≥98% (HPLC), Chemokine receptor CXCR3 agonist, powder

Synonym(s):

(3S)-N-[(1S)-5-Amino-1-[[(2,2-diphenylethyl)amino]carbonyl]pentyl]-2-(1,4-dioxo-4-phenylbutyl)-1,2,3,4-tetrahydro-3-isoquinolinecarboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
$256.00
25 mg
Check Cart for Availability
$1,020.00

About This Item

Empirical Formula (Hill Notation):
C40H44N4O4
CAS Number:
Molecular Weight:
644.80
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

$256.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

VUF10661, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

Biochem/physiol Actions

VUF10661 is a potent agonist of the chemokine receptor CXCR3.
VUF10661 is a potent agonist of the chemokine receptor CXCR3. VUF10661 induces receptor internalization, recruitment of ?-arrestins and activates Gi proteins with similar potency to endogenous CXCL ligands.

Features and Benefits

This compound is featured on the Chemokine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1107SML1781SML0789
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 15 mg/mL, clear

color

white to beige

color

white to beige

color

white to beige

color

white to beige


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters