Skip to Content
Merck
All Photos(1)

Key Documents

SML0785

Sigma-Aldrich

AZ628

≥98% (HPLC)

Synonym(s):

3-(1-Cyano-1-methylethyl)-N-[3-[(3,4-dihydro-3-methyl-4-oxo-6-quinazolinyl)amino]-4-methylphenyl]-benzamide, AZ-628

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$155.00
25 MG
$605.00

$155.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$155.00
25 MG
$605.00

About This Item

Empirical Formula (Hill Notation):
C27H25N5O2
CAS Number:
Molecular Weight:
451.52
UNSPSC Code:
12352200
NACRES:
NA.77

$155.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

[n]1([c](c2c(nc1)ccc(c2)Nc3c(ccc(c3)NC(=O)c4cc(ccc4)C(C)(C)C#N)C)=O)C

InChI

1S/C27H25N5O2/c1-17-8-9-21(31-25(33)18-6-5-7-19(12-18)27(2,3)15-28)14-24(17)30-20-10-11-23-22(13-20)26(34)32(4)16-29-23/h5-14,16,30H,1-4H3,(H,31,33)

InChI key

ZGBGPEDJXCYQPH-UHFFFAOYSA-N

Biochem/physiol Actions

AZ628 is a potent Raf kinase inhibitor with IC50 values of 105 nM for wild-type B-raf, 34 nM for Raf-B V600E and 29 nM for wild-type c-Raf-1. AZ628 is a type II inhibitor that binds to the catalytically inactive Asp-Phe-Gly (DFG)-out configuration.
AZ628 is a potent Raf kinase inhibitor.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

François Sipieter et al.
iScience, 24(9), 103074-103074 (2021-09-28)
ERK1/2 involvement in cell death remains unclear, although many studies have demonstrated the importance of ERK1/2 dynamics in determining cellular responses. To untangle how ERK1/2 contributes to two cell death programs, we investigated ERK1/2 signaling dynamics during hFasL-induced apoptosis and
Shannon M White et al.
Developmental cell, 49(3), 425-443 (2019-05-08)
Merlin/NF2 is a bona fide tumor suppressor whose mutations underlie inherited tumor syndrome neurofibromatosis type 2 (NF2), as well as various sporadic cancers including kidney cancer. Multiple Merlin/NF2 effector pathways including the Hippo-YAP/TAZ pathway have been identified. However, the molecular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service