Skip to Content
Merck
All Photos(1)

Key Documents

SML0754

Sigma-Aldrich

3-Fluoroamphetamine hydrochloride

≥98% (HPLC)

Synonym(s):

3-FA hydrochloride, 3-Fluoro-α-methyl-benzeneethanamine hydrochloride, PAL-353 hydrochloride, RS)-1-(3-Fluorophenyl)propan-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12FN · HCl
CAS Number:
Molecular Weight:
189.66
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C9H12FN.ClH/c1-7(11)5-8-3-2-4-9(10)6-8;/h2-4,6-7H,5,11H2,1H3;1H

InChI key

HKAIRPOFLITWEU-UHFFFAOYSA-N

Biochem/physiol Actions

3-Fluoroamphetamine hydrochloride is a monoamine releaser and potential designer drug derivative of amphetamine.
3-Fluoroamphetamine hydrochloride is a monoamine releaser; potential designer drug derivative of amphetamine.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Repr. 2 - STOT RE 2 - STOT SE 3

Target Organs

Brain, Central nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service