Skip to Content
Merck
All Photos(1)

Key Documents

SML0699

Sigma-Aldrich

4′-Methoxyflavone

≥97% (HPLC)

Synonym(s):

2-(4-Methoxyphenyl)-4H-chromen-4-one, 2-(4-Methoxyphenyl)chromone

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$258.00

$258.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
50 MG
$258.00

About This Item

Empirical Formula (Hill Notation):
C16H12O3
CAS Number:
Molecular Weight:
252.26
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77

$258.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Assay

≥97% (HPLC)

form

lyophilized powder

storage condition

protect from light

color

white

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[o]1c2c([c](cc1c3ccc(cc3)OC)=O)cccc2

InChI

1S/C16H12O3/c1-18-12-8-6-11(7-9-12)16-10-14(17)13-4-2-3-5-15(13)19-16/h2-10H,1H3

InChI key

OMICQBVLCVRFGN-UHFFFAOYSA-N

Biochem/physiol Actions

4-methoxyflavone is a neuroprotective agent that inhibits the accumulation of poly (ADP-ribose) and neuronal cell death following chemically-induced DNA damage or NMDA excitation.
4-methoxyflavone is a neuroprotective agent that is an inhibitor of PARP-1 mediated cell death (parthanatos).

Other Notes

Light sensitive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R N Gacche et al.
Archives of biochemistry and biophysics, 577-578, 35-48 (2015-05-06)
Relationship between structural diversity and biological activities of flavonoids has remained an important discourse in the mainstream of flavonoid research. In the current study anti-angiogenic, cytotoxic, antioxidant and cyclooxygenase (COX) inhibitory activities of diverse class of flavonoids including hydroxyl and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service