Skip to Content
Merck
All Photos(1)

Key Documents

SML0698

Sigma-Aldrich

CID 11210285 hydrochloride

≥98% (HPLC)

Synonym(s):

2-Amino-4-(3,4-(methylenedioxy)benzylamino)-6-(3-methoxyphenyl)pyrimidine hydrochloride, AMBMP, N4-(1,3-benzodioxol-5-ylmethyl)-6-(3-methoxyphenyl)-2,4-pyrimidinediamine hydrochloride, Wnt Agonist

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$150.00
25 MG
$485.00

$150.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$150.00
25 MG
$485.00

About This Item

Empirical Formula (Hill Notation):
C19H18N4O3 · HCl
CAS Number:
Molecular Weight:
386.83
UNSPSC Code:
12352200
NACRES:
NA.77

$150.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(Cc3cc4c(cc3)OCO4)c1nc(nc(c1)c2cc(ccc2)OC)N

InChI

1S/C19H18N4O3/c1-24-14-4-2-3-13(8-14)15-9-18(23-19(20)22-15)21-10-12-5-6-16-17(7-12)26-11-25-16/h2-9H,10-11H2,1H3,(H3,20,21,22,23)

InChI key

FABQUVYDAXWUQP-UHFFFAOYSA-N

Application

CID 11210285 hydrochloride has been used as a Wnt agonist.[1]

Biochem/physiol Actions

CID 11210285 is a cell-permeable, potent and selective activator of Wnt signaling without inhibiting GSK-3β.
CID 11210285 is a cell-permeable, potent and selective activator of Wnt signaling without inhibiting GSK-3β. CID 11210285is a Wnt agonist.

Other Notes

Light sensitve and air sensitive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The activation of Wnt signaling by a STAT6-dependent macrophage phenotype promotes mucosal repair in murine IBD.
Cosin-Roger J, et al.
Mucosal Immunology, 9(4), 986-986 (2016)
Amalia Rosner et al.
Developmental biology, 448(2), 320-341 (2018-11-06)
Inhibitors of Apoptosis Protein (IAP) genes participate in processes like apoptosis, proliferation, innate immunity, inflammation, cell motility, differentiation and in malignancies. Here we reveal 25 IAP genes in the tunicate Botryllus schlosseri's genome and their functions in two developmental biology

Questions

  1. What other vehicles can be used for the administration of the agonist in a mouse model? It can be administered locally (bone tissue) or only systemically?

    1 answer
    1. This product has not been tested for in vivo or in vitro applications. The method of administration will need to be validated by the end user. It may be helpful to view peer-reviewed articles. Please see the two publications below :

      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC5662016/
      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC8565963/

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service