Skip to Content
Merck
All Photos(1)

Key Documents

SML0683

Sigma-Aldrich

JNJ-40418677

≥98% (HPLC)

Synonym(s):

2-(S)-(3,5-Bis(4-(trifluoromethyl)phenyl)phenyl)-4-methylpentanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$191.00
25 MG
$758.00

$191.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$191.00
25 MG
$758.00

About This Item

Empirical Formula (Hill Notation):
C26H22F6O2
CAS Number:
Molecular Weight:
480.44
UNSPSC Code:
12352200
NACRES:
NA.77

$191.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

originator

Johnson & Johnson

relevant disease(s)

Alzheimer′s disease

storage temp.

2-8°C

Biochem/physiol Actions

JNJ-40418677 is a γ-secretase modulator that selectively blocks γ-site cleavage of the APP without affecting processing of Notch. In human neuroblastoma cells and rat primary neuronal cultures, JNJ-40418677 blocked secretion of Aβb42 (IC50 = 20 nM), with no affects on total Ab concentration, Notch signaling or COX activity. In vivo experiments showed that JNJ-40418677 dose dependently inhibited Aβ42 accumulation in the brain while promoting an increase in Aβ38.
JNJ-40418677 is a γ-secretase modulator.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 4

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service