Skip to Content
Merck
All Photos(1)

Key Documents

SML0646

Sigma-Aldrich

GMX1778

≥98% (HPLC)

Synonym(s):

CHS 828, CHS-828, GMX 1778, N-[6-(4-Chlorophenoxy)hexyl]-N′-cyano-N′′-4-pyridinyl-guanidine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$138.00
25 MG
$533.00

$138.00


Estimated to ship onMay 19, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$138.00
25 MG
$533.00

About This Item

Empirical Formula (Hill Notation):
C19H22ClN5O
CAS Number:
Molecular Weight:
371.86
UNSPSC Code:
12352200
NACRES:
NA.77

$138.00


Estimated to ship onMay 19, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

Clc1ccc(cc1)OCCCCCCN\C(=N\C#N)\Nc2ccncc2

InChI

1S/C19H22ClN5O/c20-16-5-7-18(8-6-16)26-14-4-2-1-3-11-23-19(24-15-21)25-17-9-12-22-13-10-17/h5-10,12-13H,1-4,11,14H2,(H2,22,23,24,25)

InChI key

BOIPLTNGIAPDBY-UHFFFAOYSA-N

Application

GMX1778 has been used as NAMPT inhibitor, to study its effects on nicotinamide adenine dinucleotide (NAD) contents of SH-SY5Y cells exposed to Vacor.[1]

Biochem/physiol Actions

GMX1778 (CHS-828) is a competitive inhibitor of nicotinamide phosphoribosyltransferase (NAMPT) that exhibits a potent anticancer activity both in vitro and in vivo. GMX1778 exerts a cytotoxic effect by decreasing the cellular level of NAD+. GMX1778 increases intracellular ROS in cancer cells but does not induce ROS in normal cells.
GMX1778 can regulate redox status and has the ability to simulate ROS in cancer cells.[2]
GMX1778 is a competitive inhibitor of nicotinamide phosphoribosyltransferase (NAMPT).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of nicotinamide phosphoribosyltransferase (NAMPT) activity by small molecule GMX1778 regulates reactive oxygen species (ROS)-mediated cytotoxicity in a p53-and nicotinic acid phosphoribosyltransferase1 (NAPRT1)-dependent manner
Cerna D, et al.
The Journal of Biological Chemistry, 287(26), 22408-22417 (2012)
Identification of the nicotinamide salvage pathway as a new toxification route for antimetabolites
Buonvicino D, et al.
Cell Chemical Biology, 25(4), 471-482 (2018)
Daniela Buonvicino et al.
Cell chemical biology, 25(4), 471-482 (2018-02-27)
Interest in the modulation of nicotinamide adenine dinucleotide (NAD) metabolome is gaining great momentum because of its therapeutic potential in different human disorders. Suppression of nicotinamide salvage by nicotinamide phosphoribosyl transferase (NAMPT) inhibitors, however, gave inconclusive results in neoplastic patients because
Chiara Zucal et al.
BMC cancer, 15, 855-855 (2015-11-07)
Nicotinamide phosphoribosyltransferase (NAMPT), the rate-limiting enzyme in NAD(+) biosynthesis from nicotinamide, is one of the major factors regulating cancer cells metabolism and is considered a promising target for treating cancer. The prototypical NAMPT inhibitor FK866 effectively lowers NAD(+) levels in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service