Skip to Content
Merck
All Photos(1)

Key Documents

SML0644

Sigma-Aldrich

SANT-2

≥95% (HPLC)

Synonym(s):

N-[3-(1H-Benzimidazol-2-yl)-4-chlorophenyl]-3,4,5-triethoxybenzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$162.00

$162.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$162.00

About This Item

Empirical Formula (Hill Notation):
C26H26ClN3O4
CAS Number:
Molecular Weight:
479.96
UNSPSC Code:
51111800
NACRES:
NA.77

$162.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

Clc1c(cc(cc1)NC(=O)c4cc(c(c(c4)OCC)OCC)OCC)c2[nH]c3c(n2)cccc3

InChI

1S/C26H26ClN3O4/c1-4-32-22-13-16(14-23(33-5-2)24(22)34-6-3)26(31)28-17-11-12-19(27)18(15-17)25-29-20-9-7-8-10-21(20)30-25/h7-15H,4-6H2,1-3H3,(H,28,31)(H,29,30)

InChI key

VQOJFGFKIVFMDH-UHFFFAOYSA-N

Biochem/physiol Actions

SANT-2 is a potent sonic hedgehog pathway (shh) antagonist that directly inhibits by binding to the smoothened receptor.
SANT-2 is a potent sonic hedgehog pathway (shh) antagonist,

Other Notes

Light sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service