Skip to Content
Merck
All Photos(1)

Key Documents

SML0610

Sigma-Aldrich

Nitidine chloride

≥97% (HPLC)

Synonym(s):

2,3-Dimethoxy-12-methyl-[1,3]benzodioxolo[5,6-c]phenanthridinium chloride, NSC 146397

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18NO4 · Cl
CAS Number:
Molecular Weight:
383.82
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

shipped in

wet ice

storage temp.

−20°C

SMILES string

C[N+]1=CC2=CC(OC)=C(OC)C=C2C3=CC=C4C=C5C(OCO5)=CC4=C31.[Cl-]

InChI

1S/C21H18NO4.ClH/c1-22-10-13-7-17(23-2)18(24-3)8-15(13)14-5-4-12-6-19-20(26-11-25-19)9-16(12)21(14)22;/h4-10H,11H2,1-3H3;1H/q+1;/p-1

InChI key

QLDAACVSUMUMOR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Nitidine chloride is a natural product with anti-cancer activity.
Nitidine chloride is a natural product with anti-cancer activity. Its mechanism of action likely involves several pathways. Nitidine chloride has been found to inhibit topoisomerase I and topoisomerase II, induce cell apoptosis by activation of the caspase-dependent pathway, suppress c-Src/FAK associated signaling pathways and suppress Janus kinase 2/STAT3 signaling and the expression of STAT3-dependent target genes, including cyclin D1, Bcl-xL, and VEGF. Nitidine chloride has also been found to have anti-malaria activity.

Features and Benefits

This compound is featured on the Caspases, Fak and JAKs pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ziqiang Wang et al.
Journal of ethnopharmacology, 144(1), 145-150 (2012-09-14)
Zanthoxylum nitidium (Roxb.) DC. has long been used as a traditional herbal medicine for inflammatory diseases such as rheumatic arthritis and peridentitis. However, the anti-inflammatory mechanism of Nitidine chloride has not been fully elucidated. To determine the anti-inflammatory effects and
Shu Yang et al.
Fitoterapia, 81(8), 1026-1032 (2010-07-14)
The inducing abilities of two alkaloids, Sanguinarine (San) and Nitidine (Nit), to fold human telomeric DNA d[5'-TTGGG(TTAGGG)(3)A-3'] (H24A) into G-quadruplex structures has been investigated by using CD and (1)H NMR spectroscopy. It is found that, under salt-deficiency condition, San could
S Zhang et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 24(9), 649-650 (2002-01-22)
To determine the nitidine in different parts of Zanthoxylum nitidum (Roxb.) DC. HPLC method was used. The column was HYPERSIL BDS C18. The mobile phase was 0.02 mol/L monobasic potassium phosphate: acetonitrile (66:34). The detective wavelength was 328 nm. The
Huagang Liu et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 34(11), 1406-1409 (2009-09-24)
A high performance liquid chromatography (HPLC) method was developed to determine the concentration of nitidine chloride in plasma and successfully applied to study pharmacokinetics after i.v. administration in rabbits. Twelve rabbits, randomized into 2 groups , were given i.v. at
Li-Ping Bai et al.
Bioorganic & medicinal chemistry, 14(16), 5439-5445 (2006-05-30)
A comparative study on the intercalating binding of sanguinarine, chelerythrine, and nitidine with CT DNA, poly(dG-dC).poly(dG-dC), poly(dA-dT).poly(dA-dT), and seven sequence-designed double-stranded oligodeoxynucleotides has been performed using fluorometric and spectrophotometric techniques, aiming at providing insights into their sequence selectivity for DNA-binding.

Articles

The focal adhesion kinase (FAK) is a cytoplasmic protein tyrosine kinase. FAK has been implicated as a downstream signaling molecule that functions in the control of several integrin-regulated biological processes.

Agents reported to activate cellular caspases include chemotherapeutic drugs, TNF receptor agonists, and other enzymes. Inhibitors of apoptosis were the first identified endogenous caspase inhibitors.

Src is the original member of a group of non-receptor protein-tyrosine kinases termed the Src family kinases (SFKs). Learn about the SFKs that, in humans, consist of eight members, a small group of atypical members, and two related kinases.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service