Skip to Content
Merck
All Photos(1)

Documents

SML0606

Sigma-Aldrich

UBP608

≥98% (HPLC)

Synonym(s):

6-Bromo-2-oxo-2H-chromene-3-carboxylic acid, 6-Bromo-3-carboxycoumarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5BrO4
CAS Number:
Molecular Weight:
269.05
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

InChI

1S/C10H5BrO4/c11-6-1-2-8-5(3-6)4-7(9(12)13)10(14)15-8/h1-4H,(H,12,13)/p-1

InChI key

XFQHPAXNKDYMOX-UHFFFAOYSA-M

Biochem/physiol Actions

UBP608 is NMDA receptor family allosteric modulator that selectively inhibits GluN1/GluN2A receptors with a 23-fold selectivity compared to GluN1/GluN2D receptors. It appears to distinguish between GluN2A and GluN2B, and between GluN2C and GluN2D. UBP608 does not act at either the glutamate or glycine binding sites, the ion channel, or the NTD. It appears that UBP608 is acting at the dimer interface between individual subunit ligand-binding domains.

Features and Benefits

This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to ion channel family glutamate receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service