Skip to Content
Merck
All Photos(1)

Key Documents

SML0556

Sigma-Aldrich

LY344864 hydrochloride

≥98% (HPLC)

Synonym(s):

N-[(3R)-3-(Dimethylamino)-2,3,4,9-tetrahydro-1H-carbazol-6-yl]-4-fluoro-benzamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$155.00

$155.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
5 MG
$155.00

About This Item

Empirical Formula (Hill Notation):
C21H22FN3O · HCl
CAS Number:
Molecular Weight:
387.88
UNSPSC Code:
12352200
NACRES:
NA.77

$155.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)C(=O)Nc2cc3c([nH]c4c3C[C@@H](CC4)N(C)C)cc2.Cl

InChI

1S/C21H22FN3O.ClH/c1-25(2)16-8-10-20-18(12-16)17-11-15(7-9-19(17)24-20)23-21(26)13-3-5-14(22)6-4-13;/h3-7,9,11,16,24H,8,10,12H2,1-2H3,(H,23,26);1H/t16-;/m1./s1

InChI key

OKUHLSYESBLBCP-PKLMIRHRSA-N

Biochem/physiol Actions

LY344864 hydrochloride is a potent and selective 5-HT1F receptor agonist.
LY344864 hydrochloride is a potent and selective 5-HT1F receptor agonist. It has an EC50 of 3 nM, and displays > 80-fold selectivity over other 5-HT receptors
LY344864 might be useful in the treatment approach for migraine. LY344864 is found to prevent the extravasation of dura protein and reduce c-fos like immunoreactions induced by capsaicin.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

5HT1F-and 5HT7-receptor agonists for the treatment of migraines.
Agosti R M.
CNS & Neurological Disorders Drug Targets, 6(4), 235-237 (2007)
Hans Jürgen Solinski et al.
Cell reports, 26(13), 3561-3573 (2019-03-28)
Itch is an unpleasant skin sensation that can be triggered by exposure to many chemicals, including those released by mast cells. The natriuretic polypeptide b (Nppb)-expressing class of sensory neurons, when activated, elicits scratching responses in mice, but it is

Articles

We offer many products related to serotonin receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service