Skip to Content
Merck
All Photos(1)

Key Documents

SML0544

Sigma-Aldrich

GRI977143

≥98% (HPLC)

Synonym(s):

2-[[3-(1,3-Dioxo-1H-benz[de]isoquinolin-2(3H)-yl)propyl]thio]-benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$161.00

$161.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$161.00

About This Item

Empirical Formula (Hill Notation):
C22H17NO4S
CAS Number:
Molecular Weight:
391.44
UNSPSC Code:
12352200
NACRES:
NA.77

$161.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

S(CCC[n]2[c](c3c4c([c]2=O)cccc4ccc3)=O)c1c(cccc1)C(=O)O

InChI

1S/C22H17NO4S/c24-20-16-9-3-6-14-7-4-10-17(19(14)16)21(25)23(20)12-5-13-28-18-11-2-1-8-15(18)22(26)27/h1-4,6-11H,5,12-13H2,(H,26,27)

InChI key

GMVZUCHUOYUMLL-UHFFFAOYSA-N

Biochem/physiol Actions

GRI977143 is a potent, specific LPA2 receptor agonist with no affinity for other LPA receptors. GRI977143 inhibits the initiation of apoptosis induced by adriamycin or serum withdrawal in MEFs derived from LPA2 knock-out mice transfected with an LPA2 expression vector, but has no affect in non-transfected cells.
GRI977143 is a potent, specific LPA2 receptor agonist.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sigma-Aldrich offers many products related to lysophospholipid receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service