Skip to Content
Merck
All Photos(1)

Key Documents

SML0526

Sigma-Aldrich

EHop-016

≥98% (HPLC)

Synonym(s):

N4-(9-Ethyl-9H-carbazol-3-yl)-N2-[3-(4-morpholinyl)propyl]-2,4-pyrimidinediamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$138.00
25 MG
$554.00

$138.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$138.00
25 MG
$554.00

About This Item

Empirical Formula (Hill Notation):
C25H30N6O
CAS Number:
Molecular Weight:
430.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$138.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCn1c2ccccc2c3cc(Nc4ccnc(NCCCN5CCOCC5)n4)ccc13

InChI

1S/C25H30N6O/c1-2-31-22-7-4-3-6-20(22)21-18-19(8-9-23(21)31)28-24-10-12-27-25(29-24)26-11-5-13-30-14-16-32-17-15-30/h3-4,6-10,12,18H,2,5,11,13-17H2,1H3,(H2,26,27,28,29)

InChI key

AFTZZRFCMOAFCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

EHop-016 has been used as the EHop inhibitor to study signalling diversity during Chlamydia trachomatis cell entry.[1] It has also been used as a RAC inhibitor.[2]

Biochem/physiol Actions

EHop-016 is a potent and specific inhibitor of Rac1 and Rac3 GTPase activity that inhibits Vav2 interaction with Rac, Rac-activated PAK1, lamellipodia formation, and cell migration. EHop-016 inhibits the Rac activity of MDA-MB-231 metastatic breast cancer cells and reduces Rac-directed lamellipodia formation.
EHop-016 is a potent and specific inhibitor of Rac1 and Rac3 GTPase activity.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GTP Binding Proteins (Low Molecular Weight) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bálint Tél et al.
Cellular and molecular life sciences : CMLS, 80(1), 31-31 (2023-01-08)
Thiopurine-induced acute pancreatitis (TIP) is one of the most common adverse events among inflammatory bowel disease patients treated with azathioprine (AZA), representing a significant clinical burden. Previous studies focused on immune-mediated processes, however, the exact pathomechanism of TIP is essentially
Chlamydia exploits filopodial capture and a macropinocytosis-like pathway for host cell entry
Ford C, et al.
PLoS Pathogens, 14(5), e1007051-e1007051 (2018)
Endophilin marks and controls a clathrin-independent endocytic pathway
Boucrot E, et al.
Nature, 517(7535), 460-460 (2015)
Bo Zhang et al.
Cells, 9(3) (2020-03-08)
Rho GTPases, including Rho, Cdc42, Rac and ROP subfamilies, are key signaling molecules in RNA polymerase II (Pol II) transcriptional control. Our prior work has shown that plant ROP and yeast Cdc42 GTPases similarly modulate Ser2 and Ser5 phosphorylation status
Ghulam Mohammad et al.
Molecular neurobiology, 56(12), 8643-8655 (2019-07-14)
Early activation of cytosolic NADPH oxidase-2 (Nox2) in diabetes increases retinal ROS production, damaging their mitochondria. The assembly of Nox2 holoenzyme requires activation of a small molecular weight G protein Rac1. Rac1 activation is regulated by guanine exchange factors and

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service