Skip to Content
Merck
All Photos(1)

Key Documents

SML0461

Sigma-Aldrich

PH-002

≥98% (HPLC)

Synonym(s):

4-[[4-[[2-(3,4-Dihydro-3-methyl-4-oxo-1-phthalazinyl)acetyl]amino]phenyl]methyl]-1-piperazinecarboxylic acid 1,1-dimethylethyl ester, PH 002, PH002

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$186.00
25 MG
$738.00

$186.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$186.00
25 MG
$738.00

About This Item

Empirical Formula (Hill Notation):
C27H33N5O4
CAS Number:
Molecular Weight:
491.58
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$186.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

O=C(N1CCN(CC2=CC=C(NC(CC3=NN(C)C(C4=C3C=CC=C4)=O)=O)C=C2)CC1)OC(C)(C)C

InChI

1S/C27H33N5O4/c1-27(2,3)36-26(35)32-15-13-31(14-16-32)18-19-9-11-20(12-10-19)28-24(33)17-23-21-7-5-6-8-22(21)25(34)30(4)29-23/h5-12H,13-18H2,1-4H3,(H,28,33)

InChI key

GSXXTLWPQMHHDJ-UHFFFAOYSA-N

Biochem/physiol Actions

PH-002 is an inhibitor of apolipoprotein (apo) E4 intramolecular domain interaction in neuronal cells that restores mitochondrial cytochrome c oxidase subunit 1 levels, rescues impairments of mitochondrial motility and neurite outgrowth. PH-002 is an apoE4 structure corrector that reverses the apoE4 detrimental effects.
PH-002 is an inhibitor of apolipoprotein (apo) E4 intramolecular domain interaction; apoE4 structure corrector.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service