Skip to Content
Merck
All Photos(1)

Key Documents

SML0371

Sigma-Aldrich

MLR-1023

≥98% (HPLC)

Synonym(s):

5-(3-Methylphenoxy)- 2(1H)-pyrimidinone, CP 26154, NSC 314335, Tolimidone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$123.00
25 MG
$477.00

$123.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$123.00
25 MG
$477.00

About This Item

Empirical Formula (Hill Notation):
C11H10N2O2
CAS Number:
Molecular Weight:
202.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$123.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL (clear solution)

storage temp.

room temp

SMILES string

Cc1cccc(OC2=CNC(=O)N=C2)c1

InChI

1S/C11H10N2O2/c1-8-3-2-4-9(5-8)15-10-6-12-11(14)13-7-10/h2-7H,1H3,(H,12,13,14)

InChI key

HJQILFPVRNHTIG-UHFFFAOYSA-N

Biochem/physiol Actions

MLR-1023 has a therapeutic potential to treat type 2 diabetes.[1]
MLR-1023 is a potent, orally active, allosteric activator of the non-receptor Src-related Lyn kinase that produces lasting glucose lowering, reduces HbA1c levels, and preserves pancreatic β-cells in db/db mice. MLR-1023 increased insulin receptor sensitivity. MLR-1023 (Tolimidone) is antiulcer agent.
MLR-1023 is a potent, orally active, allosteric activator of the non-receptor Src-related Lyn kinase; insulin receptor-potentiating agent; antiulcer agent.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the InsR page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael S Saporito et al.
The Journal of pharmacology and experimental therapeutics, 342(1), 15-22 (2012-04-05)
2(1H)-pyrimidinone,5-(3-methylphenoxy) (MLR-1023) is a candidate for the treatment of type 2 diabetes. The current studies were aimed at determining the mechanism by which MLR-1023 mediates glycemic control. In these studies, we showed that MLR-1023 reduced blood glucose levels without increasing
Fei Zhuang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(10), 13586-13596 (2020-08-29)
Endothelial microparticles (EMPs) are involved in various cardiovascular pathologies and play remarkable roles in communication between endothelial cells (ECs), which are constantly exposed to mechanical cyclic stretch (CS) following blood pressure. However, the roles of EMPs induced by CS in

Articles

Src is the original member of a group of non-receptor protein-tyrosine kinases termed the Src family kinases (SFKs). Learn about the SFKs that, in humans, consist of eight members, a small group of atypical members, and two related kinases.

We offers many products related to InsR for your research needs.

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service