Skip to Content
Merck
All Photos(1)

Key Documents

SML0362

Sigma-Aldrich

NPS2143 hydrochloride

≥98% (HPLC)

Synonym(s):

2-Chloro-6-[(2R)-3-[[1,1-dimethyl-2-(2-naphthalenyl)ethyl]amino]-2-hydroxypropoxy]-benzonitrile hydrochloride (1:1), NPS-2143 hydrochloride, SB-262470-A hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$198.00
25 MG
$792.00

$198.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$198.00
25 MG
$792.00

About This Item

Empirical Formula (Hill Notation):
C24H25ClN2O2 · HCl
CAS Number:
Molecular Weight:
445.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$198.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]20/D +20 to +24°, c = 1.0 in methanol

storage condition

desiccated

color

white to beige

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Cl.CC(C)(Cc1ccc2ccccc2c1)NC[C@@H](O)COc3cccc(Cl)c3C#N

InChI

1S/C24H25ClN2O2.ClH/c1-24(2,13-17-10-11-18-6-3-4-7-19(18)12-17)27-15-20(28)16-29-23-9-5-8-22(25)21(23)14-26;/h3-12,20,27-28H,13,15-16H2,1-2H3;1H/t20-;/m1./s1

InChI key

ZEBNDUQLNGYBNL-VEIFNGETSA-N

Application

NPS2143 hydrochloride has been used to study the function of calcium-sensing receptor :
  • In the regulation of intestinal calcium transport.[1]
  • In the regulation of local negative feedback of intestinal calcium absorption.[2]
  • In sensorimotor decision making during development.[3]

Biochem/physiol Actions

NPS2143 is a phenylalkylamine calcilytic compound,[4] and might be useful in treating autosomal dominant hypocalcemia.[5] It serves as a blocking agent for hERG (human ether-a-go-go-related gene ) channel, CYP2D6 (cytochrome P450 2D6 ) and monoamine transporters.[6]
NPS2143 is a selective calcium-sensing receptor (CaR) antagonist.
NPS2143 is a selective calcium-sensing receptor (CaR) antagonist. The compound blocks increases in cytoplasmic calcium concentrations with an IC50 of 43 nM, and stimulates parathyroid hormone (PTH) secretion from bovine parathyroid cells over a wide range of extracellular calcuim concentrations.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fen-Fen Liu et al.
Cardiorenal medicine (2022-10-11)
Introduction Apoptosis of vascular smooth muscle cells induced by hyperphosphatemia is a critical mechanism of chronic kidney disease-related vascular disorders. The present study investigated whether extracellular calcium-sensing receptor (CaSR) regulates stanniocalcin 2 (STC2) expression in HAoSMCs in vitro and whether
The calcilytic agent NPS 2143 rectifies hypocalcemia in a mouse model with an activating calcium-sensing receptor (CaSR) mutation: relevance to autosomal dominant hypocalcemia type 1 (ADH1)
Hannan FM, et al.
Endocrinology, 156(9), 3114-3121 (2015)
A Forward Genetic Screen in Zebrafish Identifies the G-Protein-Coupled Receptor CaSR as a Modulator of Sensorimotor Decision Making
Jain RA, et al.
Current Biology, 28(9), 1357-1369 (2018)
Activation of calcium-sensing receptor by allosteric agonists cinacalcet and AC-265347 abolishes the 1, 25 (OH) 2D3-induced Ca2+ transport: Evidence that explains how the intestine prevents excessive Ca2+ absorption
Wongdee K, et al.
Archives of Biochemistry and Biophysics, 657, 15-22 (2018)
Progress in Medicinal Chemistry, 57 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service