Skip to Content
Merck
All Photos(1)

Documents

SML0347

Sigma-Aldrich

MJC13

≥98% (HPLC)

Synonym(s):

N-(2,3-dichlorophenyl)-cyclohexanecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15Cl2NO
CAS Number:
Molecular Weight:
272.17
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

Clc1cccc(NC(=O)C2CCCCC2)c1Cl

InChI

1S/C13H15Cl2NO/c14-10-7-4-8-11(12(10)15)16-13(17)9-5-2-1-3-6-9/h4,7-9H,1-3,5-6H2,(H,16,17)

InChI key

ZYPWKRVXNGLEMU-UHFFFAOYSA-N

Biochem/physiol Actions

MJC13 inhibits the positive regulation of androgen receptor (AR) signaling by the cochaperone protein FKBP52. The molecule inhibits hormone-induced dissociation of the FKBP52, nuclear translocation of AR and AR dependent gene expression. MJC13 inhibits proliferation of the androgen-dependent prostate cancer cell lines LNCaP and LAPC4.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayesha A Shafi et al.
Steroids, 78(6), 548-554 (2013-02-06)
Androgen ablation therapy is the most common treatment for advanced prostate cancer (PCa), but most patients will develop castration-resistant prostate cancer (CRPC), which has no cure. CRPC is androgen-depletion resistant but androgen receptor (AR) dependent. AR is a nuclear receptor
Su Liang et al.
American journal of modern chromatography, 1(1), 1-11 (2015-01-17)
MJC13 is a novel molecule that has potential use for the treatment of hormone refractory prostate cancer (HRPC). The purpose of this work was to develop a liquid chromatography-tandem mass spectrometry (LC-MS/MS) method for quantification of MJC13. Itraconazole was used
Ji Ho Suh et al.
PloS one, 10(9), e0137103-e0137103 (2015-09-04)
The androgen receptor (AR) surface-directed antagonist MJC13 inhibits AR function and proliferation of prostate cancer (PC) cells. These effects are related to arrest of an AR/chaperone complex in the cytoplasm. Here, we compared MJC13 and classic AR antagonists such as
Keisuke Maeda et al.
Molecular oncology (2021-06-01)
The growth of prostate cancer is dependent on the androgen receptor (AR), which serves as a ligand-specific transcription factor. Although two immunophilins, FKBP51 and FKBP52, are known to regulate AR activity, the precise mechanism remains unclear. We found that depletion
Johanny Tonos De Leon et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(29), 11878-11883 (2011-07-07)
Drugs that target novel surfaces on the androgen receptor (AR) and/or novel AR regulatory mechanisms are promising alternatives for the treatment of castrate-resistant prostate cancer. The 52 kDa FK506 binding protein (FKBP52) is an important positive regulator of AR in

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service