Skip to Content
Merck
All Photos(1)

Key Documents

SML0266

Sigma-Aldrich

AA 29504

≥98% (HPLC)

Synonym(s):

N-[2-Amino-4-[[(2,4,6-trimethylphenyl)methyl]amino]phenyl]-carbamic acid-ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$419.00

$419.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$419.00

About This Item

Empirical Formula (Hill Notation):
C19H25N3O2
CAS Number:
Molecular Weight:
327.42
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$419.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥2 mg/mL

storage temp.

−20°C

SMILES string

CCOC(=O)Nc1ccc(NCc2c(C)cc(C)cc2C)cc1N

InChI

1S/C19H25N3O2/c1-5-24-19(23)22-18-7-6-15(10-17(18)20)21-11-16-13(3)8-12(2)9-14(16)4/h6-10,21H,5,11,20H2,1-4H3,(H,22,23)

InChI key

SCOOTUMXCXKEAD-UHFFFAOYSA-N

Biochem/physiol Actions

AA29504 is a GABAA positive modulator with a preference for extra-synaptic α4β3δ over α1β3γ2s synaptic receptors. AA29504 exhibits anxiolytic effects and reduces motor coordination in rat models.
AA29504 is a GABAA positive modulator.

Features and Benefits

This compound is featured on the GABAA Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina B Falk-Petersen et al.
Basic & clinical pharmacology & toxicology, 121(2), 119-129 (2017-03-17)
δ-Containing GABAA receptors are located extrasynaptically and mediate tonic inhibition. Their involvement in brain physiology positions them as interesting drug targets. There is thus a continued interest in establishing reliable recombinant expression systems for δ-containing GABAA receptors. Inconveniently, the recombinant

Articles

We offer many products related to GABAA receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service