Skip to Content
Merck
All Photos(1)

Key Documents

SML0252

Sigma-Aldrich

Anisodamine

≥98% (HPLC)

Synonym(s):

α-(Hydroxymethyl)-benzeneacetic acid 6-hydroxy-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, 6β-Hydroxyhyoscyamine, 6-Hydroxyhyoscyamine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$124.00
50 MG
$479.00

$124.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$124.00
50 MG
$479.00

About This Item

Empirical Formula (Hill Notation):
C17H23NO4
CAS Number:
Molecular Weight:
305.37
UNSPSC Code:
12352116
NACRES:
NA.77

$124.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: ≥5 mg/mL

storage temp.

room temp

SMILES string

CN1[C@H]2C(O)C[C@@H]1C[C@H](OC(C(CO)C3=CC=CC=C3)=O)C2

InChI

1S/C17H23NO4/c1-18-12-7-13(9-15(18)16(20)8-12)22-17(21)14(10-19)11-5-3-2-4-6-11/h2-6,12-16,19-20H,7-10H2,1H3/t12-,13-,14+,15+,16-/m0/s1

InChI key

WTQYWNWRJNXDEG-RBZJEDDUSA-N

General description

Racemic mixture of diastereomers.

Biochem/physiol Actions

Anisodamine is a muscarinic receptor antagonist. Anisodamine is a naturally occurring atropine derivative isolated from Scopolia Tangutica Maxim plant indigenous to Tibet.
Anisodamine is a muscarinic receptor antagonist; anticholinergic.
Anisodamine is a non‐specific cholinergic antagonist. It is considered less efficient and less toxic than atropine. Anisodamine interferes with liposome structure and affects cell membrane. It might act as an anti‐oxidant, in protecting against the damage caused by free radicals. Anisodamine is known to reduce cardiac conduction and also prevents arrhythmia. It can block thromboxane synthesis and might possess anti‐thrombotic function. Anisodamine has been useful in a number therapies including septic shock, circulatory disorders, organophosphorus poisoning, opiate addiction, snake bite and radiation damage. Disorders, such as migraine, gastric ulcers, rheumatoid arthritis, gastrointestinal colic, eclampsia, respiratory diseases, acute glomerular nephritis and obstructive jaundice can be treated with the help of anisodamine.[1]

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The pharmacological properties of anisodamine
Poupko JM, et al.
Journal of Toxicology, 27(2), 116-121 (2007)

Articles

Muscarinic acetylcholine receptors mediate acetylcholine actions in CNS and non-nervous tissues, crucial for cell signaling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service