Skip to Content
Merck
All Photos(1)

Key Documents

SML0210

Sigma-Aldrich

BMS-299897

≥98% (HPLC)

Synonym(s):

2-[(1R)-1-[[(4-Chlorophenyl)sulfonyl](2,5-difluorophenyl)amino]ethyl]-5-fluoro-benzenebutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21ClF3NO4S
CAS Number:
Molecular Weight:
511.94
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥20 mg/mL

storage temp.

−20°C

SMILES string

Fc1c(cc(cc1)F)N([S](=O)(=O)c3ccc(cc3)Cl)[C@H](C)c2c(cc(cc2)F)CCCC(=O)O

InChI

1S/C24H21ClF3NO4S/c1-15(21-11-7-18(26)13-16(21)3-2-4-24(30)31)29(23-14-19(27)8-12-22(23)28)34(32,33)20-9-5-17(25)6-10-20/h5-15H,2-4H2,1H3,(H,30,31)/t15-/m1/s1

InChI key

IZAOBRWCUGOKNH-OAHLLOKOSA-N

Application

BMS-299897 has been used as an γ-secretase inhibitor to stop high glucose (HG) mediated amyloid β-peptide (Aβ) secretion.

Biochem/physiol Actions

BMS-299897 is a gamma-secretase inhibitor
BMS-299897 is a potent inhibitor of γ-secretase. The comound is 15-fold more selective for inhibiting the cleavage of β-amyloid precursor protein over Notch cleavage (IC50s 7.1 and 105.9, respectively). In vivo, BMS-299897 blocks the formation of Aβ40 and Aβ42 in the brain and CSF without affecting maturation of CD8+ T cells or intestinal goblet cells, suggesting Notch signalling was not significantly inhibited.
BMS-299897 may be used as a therapeutic for Alzheimer′s disease.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A-Ching Chao et al.
CNS neuroscience & therapeutics, 22(4), 291-297 (2016-02-05)
Amyloid beta-peptide (Aβ), the main component of senile plaques in the Alzheimer's disease (AD) brains, is generated from sequential cleavage of amyloid precursor protein (APP) by β- and γ-secretase. Hyperglycemia in diabetes may compromise barrier integrity in endothelial cells (ECs).
Donglu Zhang et al.
Current drug metabolism, 7(8), 883-896 (2006-12-16)
BMS-299897 is a gamma-secretase inhibitor that has the potential for treatment of Alzheimer's disease. The metabolism of [(14)C]BMS-299897 was investigated in human liver microsomes, in rat, dog, monkey and human hepatocytes and in bile duct cannulated rats. Seven metabolites (M1-M7)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service