Skip to Content
Merck
All Photos(1)

Key Documents

SML0206

Sigma-Aldrich

EAA-090

≥98% (HPLC)

Synonym(s):

P-[2-(8,9-dioxo-2,6-diazabicyclo[5.2.0]non-1(7)-en-2-yl)ethyl]-phosphonic acid, WAY-126090

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N2O5P
CAS Number:
Molecular Weight:
260.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

color

light yellow to yellow

solubility

H2O: ≥2 mg/mL

originator

Wyeth

storage temp.

2-8°C

SMILES string

OP(O)(=O)CCN1CCCNC2=C1C(=O)C2=O

InChI

1S/C9H13N2O5P/c12-8-6-7(9(8)13)11(3-1-2-10-6)4-5-17(14,15)16/h10H,1-5H2,(H2,14,15,16)

InChI key

BDABGOLMYNHHTR-UHFFFAOYSA-N

Application

EAA-090 was used to study the role of glutamate receptor-mediated signaling in the formation of filipodia and secondary dendrites in rat hippocampal neurons.

Biochem/physiol Actions

EAA-090 inhibits the binding of ligands to glutamate receptors and blocks NMDA-induced currents. It is neuroprotective against glutamate and ischemia-induced toxicity.
EAA-090 is a potent NMDA receptor antagonist with neuroprotective properties. In isoflurane-anesthetized dogs, EAA-090 reduces the minimum alveolar concentration (MAC) of isoflurane and increases systolic, mean, and diastolic arterial blood pressure.

Features and Benefits

This compound was developed by Wyeth. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucy Sun et al.
The Journal of pharmacology and experimental therapeutics, 310(2), 563-570 (2004-04-13)
Two novel N-methyl-d-aspartate (NMDA) antagonists with unique chemical structures, EAA-090 (2-[8,9-dioxo-2, 6-diazabicyclo[5.2.0]non-1(7)-en2-yl]ethylphosphonic acid) and EAB-318 (R-alpha-amino-5-chloro-1-(phosphonomethyl)-1H-benzimidazole-2-propanoic acid hydrochloride), were compared with CGS-19755 (Selfotel) in ligand binding, electrophysiology, and neuroprotection assays. CGS-19755, EAA-090 and EAB-318 inhibited [(3)H]3-(2-carboxypiperazin-4-yl)propyl-1-phosphonic acid binding to NMDA
Frank Henle et al.
Naunyn-Schmiedeberg's archives of pharmacology, 385(2), 171-180 (2011-10-29)
GluN receptors are heteromers of obligatory GluN1 subunits and GluN2(A-D) subunits. In the present study, we addressed the question whether GluN2A and GluN2B subunits play distinct roles in the formation of filopodia and dendrites during the early development of hippocampal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service