Skip to Content
Merck
All Photos(1)

Key Documents

SML0129

Sigma-Aldrich

AC-265347

≥98% (HPLC)

Synonym(s):

1-(1,3-benzothiazol-2-yl)-1-(2,4-dimethylphenyl)ethanol, a-(2,4-Dimethylphenyl)-a-methyl-2-benzothiazolemethanol.

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$141.00
25 MG
$456.00

$141.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$141.00
25 MG
$456.00

About This Item

Empirical Formula (Hill Notation):
C17H17NOS
CAS Number:
Molecular Weight:
283.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$141.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥19 mg/mL

storage temp.

2-8°C

SMILES string

Cc1ccc(c(C)c1)C(C)(O)c2nc3ccccc3s2

InChI

1S/C17H17NOS/c1-11-8-9-13(12(2)10-11)17(3,19)16-18-14-6-4-5-7-15(14)20-16/h4-10,19H,1-3H3

InChI key

IGSZVEPQZANNAB-UHFFFAOYSA-N

Application

AC-265347 may be used in calcium-sensing receptor-mediated signaling.

Biochem/physiol Actions

AC-265347 is a calcimimetic that acts as agonist to calcium-sensing receptor. It reduces serum parathyroid hormone and plasma ionizable calcium.[1][2]
AC-265347 is a human calcium-sensing receptor (CaSR) allosteric agonist. AC-265347 activates CaSR signaling in cellular proliferation and phosphatidyl inositol (PI) hydrolysis assays.
AC-265347 is a human calcium-sensing receptor (CaSR) allosteric agonist; calcimimetic

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pablo A Ureña Torres et al.
Kidney international, 82(1), 19-25 (2012-03-23)
Renal function impairment goes along with a disturbed calcium, phosphate, and vitamin D metabolism, resulting in secondary hyperparathyroidism (sHPT). These mineral metabolism disturbances are associated with soft tissue calcifications, particularly arteries, cardiac valves, and myocardium, ultimately associated with increased risk
Jian-Nong Ma et al.
The Journal of pharmacology and experimental therapeutics, 337(1), 275-284 (2011-01-18)
We discovered structurally novel human calcium-sensing receptor (CaSR) allosteric agonists and compared their pharmacology to phenylalkylamine calcimimetics. 1-Benzothiazol-2-yl-1-(2,4-dimethyl-phenyl)-ethanol (AC-265347) activated CaSR signaling in cellular proliferation and phosphatidylinositol (PI) hydrolysis assays with potencies of 30 and 10 nM, respectively. (S)-1-Benzothiazol-2-yl-1-(2,4-dimethyl-phenyl)-ethanol) [(S)-AC-265347]
Shane D Hellyer et al.
Molecular pharmacology, 93(5), 504-514 (2018-03-09)
Numerous positive and negative allosteric modulators (PAMs and NAMs) of class C G protein-coupled receptors (GPCRs) have been developed as valuable preclinical pharmacologic tools and therapeutic agents. Although many class C GPCR allosteric modulators have undergone subtype selectivity screening, most

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service