Skip to Content
Merck
All Photos(1)

Key Documents

SML0127

Sigma-Aldrich

Longdaysin

≥98% (HPLC)

Synonym(s):

9-isopropyl-N-(3-(trifluoromethyl)benzyl)-9H-purin-6-amine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$163.00
25 MG
$480.00

$163.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$163.00
25 MG
$480.00

About This Item

Empirical Formula (Hill Notation):
C16H16F3N5
CAS Number:
Molecular Weight:
335.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$163.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥15 mg/mL

storage temp.

2-8°C

SMILES string

CC(C)N(C=N1)C2=C1C(NCC3=CC(C(F)(F)F)=CC=C3)=NC=N2

InChI

1S/C16H16F3N5/c1-10(2)24-9-23-13-14(21-8-22-15(13)24)20-7-11-4-3-5-12(6-11)16(17,18)19/h3-6,8-10H,7H2,1-2H3,(H,20,21,22)

InChI key

REKSFCCYDQMSIN-UHFFFAOYSA-N

Application

Longdaysin may be used to study cell signaling that regulate the circadian clock and behavior.

Biochem/physiol Actions

Longdaysin inhibits CK1, ERK2, and CDK7 kinases in low mM concentrations. The activity of longdaysin results in a lengthening of circadian periods in cell based and in vivo systems. Transgenic zebrafish harboring a per3-luc reporter gene displayed a 10 hour lengthening of circadian period when treated with longdaysin. Longdaysin reduces the phosphorylation of PER1 the regulator of molecular clock function and manipulates the circadian clock.[1][2]

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsuyoshi Hirota et al.
Science (New York, N.Y.), 337(6098), 1094-1097 (2012-07-17)
Impairment of the circadian clock has been associated with numerous disorders, including metabolic disease. Although small molecules that modulate clock function might offer therapeutic approaches to such diseases, only a few compounds have been identified that selectively target core clock
Urs Albrecht
Handbook of experimental pharmacology, (217)(217), 227-239 (2013-04-23)
Circadian clocks are present in nearly all tissues of an organism, including the brain. The brain is not only the site of the master coordinator of circadian rhythms located in the suprachiasmatic nuclei (SCN) but also contains SCN-independent oscillators that
Cyclic uniaxial mechanical load enhances chondrogenesis through entraining the molecular circadian clock.
V?!g??, et al.
Journal of Pineal Research, 73, e12827-e12827 (2022)
M Abdulhadi Alagha et al.
Cartilage, 13(2_suppl), 53S-67S (2020-02-16)
Circadian rhythms in cartilage homeostasis are hypothesized to temporally segregate and synchronize the activities of chondrocytes to different times of the day, and thus may provide an efficient mechanism by which articular cartilage can recover following physical activity. While the
Sandipan Ray et al.
Life science alliance, 2(6) (2019-12-04)
Determining the exact targets and mechanisms of action of drug molecules that modulate circadian rhythms is critical to develop novel compounds to treat clock-related disorders. Here, we have used phenotypic proteomic profiling (PPP) to systematically determine molecular targets of four

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service