Skip to Content
Merck
All Photos(1)

Key Documents

SML0098

Sigma-Aldrich

Pinostilbene hydrate

≥95% (HPLC)

Synonym(s):

3,4′-Dihydroxy-5-methoxy-trans-stilbene hydrate, 3-[(1E)-2-(4-hydroxyphenyl)ethenyl]-5-methoxyphenol hydrate, trans-Pinostilbene hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$151.00
25 MG
$466.00

$151.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$151.00
25 MG
$466.00

About This Item

Empirical Formula (Hill Notation):
C15H14O3 · xH2O
CAS Number:
Molecular Weight:
242.27 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$151.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥13 mg/mL

storage temp.

2-8°C

SMILES string

O.COc1cc(O)cc(\C=C\c2ccc(O)cc2)c1

InChI

1S/C15H14O3.H2O/c1-18-15-9-12(8-14(17)10-15)3-2-11-4-6-13(16)7-5-11;/h2-10,16-17H,1H3;1H2/b3-2+;

InChI key

XZMDLGKACFLROY-SQQVDAMQSA-N

Biochem/physiol Actions

Pinostilbene is a resveratrol derivative that induced apoptosis in several cancer cell lines.
Resveratrol prodrug.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

You Chul Chung et al.
International journal of molecular sciences, 21(13) (2020-07-08)
Melanin protects our skin from harmful ultraviolet (UV) radiation. However, when produced in excess, it can cause hyperpigmentation disorders, such as melanoma, freckles, lentigo, and blotches. In this study, we investigated the effects of pinostilbene hydrate (PH) on melanogenesis. We
Caroline M Low et al.
Scientific reports, 7, 44449-44449 (2017-03-18)
The pro-inflammatory mediator leukotriene B
Md Ataur Rahman et al.
Biochimica et biophysica acta, 1861(2), 23-36 (2016-11-07)
Drug resistance from apoptosis is a challenging issue with different cancer types, and there is an interest in identifying other means of inducing cytotoxicity. Here, treatment of neuroblastoma cells with oxyresveratrol (OXYRES), a natural antioxidant, led to dose-dependent cell death

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service