Skip to Content
Merck
All Photos(1)

Key Documents

SML0069

Sigma-Aldrich

NSC 33994

≥98% (HPLC)

Synonym(s):

(E)-4,4′-(1,2-Diethyl-1,2-ethenediyl)bis[2-[(diethylamino)methyl]-phenol (9CI), 4,4′-(3E)-Hex-3-ene-3,4-diylbis{2-[(diethylamino)methyl]phenol}, G6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H42N2O2
CAS Number:
Molecular Weight:
438.65
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥25 mg/mL

storage temp.

2-8°C

SMILES string

CCN(CC)Cc1cc(ccc1O)C(\CC)=C(/CC)c2ccc(O)c(CN(CC)CC)c2

InChI

1S/C28H42N2O2/c1-7-25(21-13-15-27(31)23(17-21)19-29(9-3)10-4)26(8-2)22-14-16-28(32)24(18-22)20-30(11-5)12-6/h13-18,31-32H,7-12,19-20H2,1-6H3/b26-25+

InChI key

QFNJFVBKASKGEU-OCEACIFDSA-N

Application

NSC 33994 may be used in Jak2-mediated cell signaling studies.

Biochem/physiol Actions

NSC 33994 is a potent and specific Jak2 inhibitor that inhibits a notorious mutation, Jak2-V617F, found in large number of myeloproliferative neoplasms.
Potent and specific Jak2 inhibitor effective against Jak2-V617F mutation

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the JAKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuehui Wang et al.
Methods (San Diego, Calif.), 173, 32-43 (2019-06-22)
Influx evidences show that red long non-coding RNAs (lncRNAs) play important roles in various critical biological processes, and they afffect the development and progression of various human diseases. Therefore, it is necessary to precisely identify the lncRNA-disease associations. The identification
S Khademvatan et al.
Journal of helminthology, 93(2), 131-138 (2018-04-03)
Cystic echinococcosis (CE) caused by Echinococcus granulosus sensu lato (s.l.) is a significant zoonosis, especially in developing countries of the Middle East, with many studies focusing on CE genotypes in Iran. We performed a systematic review to determine the exact
Zhen Wang et al.
Scientific reports, 8(1), 9509-9509 (2018-06-24)
The exon junction complex (EJC) deposited on spliced mRNAs, plays a central role in the post-transcriptional gene regulation and specific gene expression. The EJC core complex is associated with multiple peripheral factors involved in various post-splicing events. Here, using recombinant
Juliet Chepngeno et al.
Veterinary research, 50(1), 84-84 (2019-10-24)
Rotavirus C (RVC) has been detected increasingly in humans and swine in different countries, including the US. It is associated with significant economic losses due to diarrheal disease in nursing piglets. In this study we aimed: (1) to determine the
Kimberly McCoy et al.
ACS nano, 12(4), 3541-3550 (2018-03-21)
Bottom-up construction of mesoscale materials using biologically derived nanoscale building blocks enables engineering of desired physical properties using green production methods. Virus-like particles (VLPs) are exceptional building blocks due to their monodispersed sizes, geometric shapes, production ease, proteinaceous composition, and

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service