Skip to Content
Merck
All Photos(1)

Key Documents

SMB01071

Sigma-Aldrich

Vaccinoside

≥90% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$459.00

$459.00


Estimated to ship onMay 02, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$459.00

About This Item

Empirical Formula (Hill Notation):
C25H28O13
CAS Number:
Molecular Weight:
536.48
UNSPSC Code:
12352205
NACRES:
NA.25

$459.00


Estimated to ship onMay 02, 2025


Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

536.48

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]2OC=C([C@@H]3[C@H]2[C@@](C=C3)(O)COC(=O)\C=C\c4ccc(cc4)O)C(=O)O

InChI

1S/C25H28O13/c26-9-16-19(29)20(30)21(31)24(37-16)38-23-18-14(15(10-35-23)22(32)33)7-8-25(18,34)11-36-17(28)6-3-12-1-4-13(27)5-2-12/h1-8,10,14,16,18-21,23-24,26-27,29-31,34H,9,11H2,(H,32,33)/b6-3+/t14-,16-,18-,19-,20+,21-,23+,24+,25+/m1/s1

InChI key

WJPRKOJNQIZCGY-OZOLURDPSA-N

General description

Vaccinoside, an iridoid glycoside, is a bioactive natural compound derived from Vaccinium bracteatum, Andromeda polifolia, and Craibiodendron henryi. Current research indicates that this metabolite is an inhibitor and may possess diverse biological activities, including antioxidant and anticancer properties.

Application

Vaccinoside is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service