Skip to Content
Merck
All Photos(1)

Key Documents

SMB01047

Sigma-Aldrich

Hexamethylquercetagetin

≥90% (LC/MS-ELSD)

Synonym(s):

Hexamethoxyflavone, Oxyayanin B trimethyl ether, Quercetagetin hexamethyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$366.00

$366.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$366.00

About This Item

Empirical Formula (Hill Notation):
C21H22O8
CAS Number:
Molecular Weight:
402.39
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

$366.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

402.39

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

[o]1c2c([c](c(c1c3cc(c(cc3)OC)OC)OC)=O)c(c(c(c2)OC)OC)OC

InChI

1S/C21H22O8/c1-23-12-8-7-11(9-13(12)24-2)18-21(28-6)17(22)16-14(29-18)10-15(25-3)19(26-4)20(16)27-5/h7-10H,1-6H3

InChI key

CHXSDKWBSFDZEU-UHFFFAOYSA-N

General description

Hexamethylquercetagetin, a polyketide-flavonoid, is a natural product commonly found in plants Jasonia montana, Pallenis spinosa, Callicarpa pilosissima, and Citrus sp. This secondary metabolite has been shown to have various biological activities, including antioxidant, anti-inflammatory, and anticancer properties.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service