Skip to Content
Merck
All Photos(1)

Key Documents

SMB00918

Sigma-Aldrich

Arabinofuranose

≥95% (HPLC)

Synonym(s):

L-Arabinofuranose, (3R,4R,5S)-5-(hydroxymethyl)oxolane-2,3,4-triol

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$202.00

$202.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$202.00

About This Item

Empirical Formula (Hill Notation):
C5H10O5
Molecular Weight:
150.13
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

$202.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

color

off-white

mp

118—121 °C

storage temp.

2-8°C

SMILES string

O1[C@H]([C@@H]([C@H](C1O)O)O)CO

InChI

1S/C5H10O5/c6-1-2-3(7)4(8)5(9)10-2/h2-9H,1H2/t2-,3-,4+,5?/m0/s1

InChI key

HMFHBZSHGGEWLO-HWQSCIPKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Arabinofuranose is a fundamental pentose sugar with a five-membered ring structure, serving as the mirror-image enantiomer to D-arabinofuranose. It falls within the class of organic compounds known as pentoses, which consist of monosaccharides featuring a carbohydrate structure composed of five carbon atoms.In biochemistry, L-Arabinofuranose plays a vital role in the synthesis of D-ribose, a key molecule in various biochemical pathways, including metabolomics. Additionally, L-Arabinofuranose is abundantly found in the cell walls and intercellular matrix polysaccharides of higher plant species, such as arabinofuranans, highlighting its significance in plant biology and the structural components of plant cells. In biochemical and cell culture studies, L-arabinofuranose can be utilized to investigate a range of biological processes, enabling the exploration of plant cell wall composition, protein glycosylation, and disease modeling, which are essential in metabolomics and drug development research.

Application

L-Arabinofuranose finds application in metabolomics, biochemical, and drug discovery research.

Features and Benefits

High quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Drugs, their targets and the nature and number of drug targets.
Imming, et al.
Nature Reviews Drug Discovery, 5, 821-834 (2007)
John P Overington et al.
Nature reviews. Drug discovery, 5(12), 993-996 (2006-12-02)
For the past decade, the number of molecular targets for approved drugs has been debated. Here, we reconcile apparently contradictory previous reports into a comprehensive survey, and propose a consensus number of current drug targets for all classes of approved

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service