Skip to Content
Merck
All Photos(1)

Key Documents

SMB00708

Sigma-Aldrich

Lysobactin

≥97% (HPLC)

Synonym(s):

Katanosin B, L-Serine, D-leucyl-L-leucyl-(βR)-β-hydroxy-L-phenylalanyl-(3R)-3-hydroxy-L-leucyl-L-leucyl-D-arginyl-L-isoleucyl-L-allothreonylglycyl-(3S)-3-hydroxy-L-asparaginyl-, (11→3)-lactone

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$175.00

$175.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$175.00

About This Item

Empirical Formula (Hill Notation):
C58H97N15O17 · xC2H4O2
CAS Number:
Molecular Weight:
1276.48 (free base basis)
UNSPSC Code:
41116107
NACRES:
NA.76

$175.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

powder

composition

counterion (acetate salt)

solubility

DMSO: 1 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

−20°C

SMILES string

N1[C@H](C(=O)NCC(=O)N[C@H](C(=O)N[C@H](C(=O)OC([C@@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@@H](C(=O)N[C@H](C1=O)[C@H](CC)C)CCCN=C(N)N)CC(C)C)[C@H](O)C(C)C)NC(=O)[C@@H](NC(=O)[C@H](N)CC(C)C)CC(C)C)c2ccccc2)CO)[C@H](O)C(=O)N)[C@@H](O)C.OC(=O)C

InChI

1S/C58H97N15O17.C2H4O2/c1-12-30(10)39-53(85)71-40(31(11)75)52(84)64-24-38(76)69-42(45(78)47(60)79)55(87)68-37(25-74)57(89)90-46(32-17-14-13-15-18-32)43(73-51(83)36(23-28(6)7)66-48(80)33(59)21-26(2)3)56(88)72-41(44(77)29(8)9)54(86)67-35(22-27(4)5)50(82)65-

InChI key

TXXDLVCKZNQGAV-QPXZYOHJSA-N

Biochem/physiol Actions

Lysobactin is a cyclic peptide with strong antibacterial activity against many bacterial strains including Staphylococcus aureus, Salmonella typhosa, and Klebsiella aerogenes. Lysobactin is reported to have MIC values of 0.2, 25, and 25 μg/mL for the above listed bacteria respectively, which is significantly higher than the potency reported for vancomycin for the same bacteria (MIC values 0.8, >100, and >100 μg/mL respectively).[1]

The mechanism of action of lysobactin is attributed to its interaction with lipid I and lipid II in the bacterial cell wall. Lysobactin forms a 1:1 complex with lipid I and lipid II causing interference in peptidoglycan synthesis which is essential for bacterial viability.[2] Furthermore, lysobactin was not found to be hemolytic at concentrations above its MIC.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D P Bonner et al.
The Journal of antibiotics, 41(12), 1745-1751 (1988-12-01)
Lysobactin, an antibiotic isolated from a strain of Lysobacter, is 2 to 4-fold more active than vancomycin against aerobic and anaerobic Gram-positive bacteria. Included in the spectrum of lysobactin are Staphylococci, Streptococci, corynebacteria, clostridia and various other Gram-positive anaerobic bacteria.
Wonsik Lee et al.
Journal of the American Chemical Society, 138(1), 100-103 (2015-12-20)
Lysobactin, also known as katanosin B, is a potent antibiotic with in vivo efficacy against Staphylococcus aureus and Streptococcus pneumoniae. It was previously shown to inhibit peptidoglycan (PG) biosynthesis, but its molecular mechanism of action has not been established. Using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service