Skip to Content
Merck
All Photos(1)

Key Documents

SMB00706

Sigma-Aldrich

Lycopene

≥98% (HPLC), from tomato

Synonym(s):

ψ,ψ-Carotene, 2,6,10,14,19,23,27,31-Octamethyl-dotriaconta-2,6,8,10,12,14,16,18,20,22,24,26,30-tridecaene

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$478.00
10 MG
$1,430.00
5 MG
$1,440.00

$478.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$478.00
10 MG
$1,430.00
5 MG
$1,440.00

About This Item

Empirical Formula (Hill Notation):
C40H56
CAS Number:
Molecular Weight:
536.87
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.28

$478.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

tomato

Quality Level

Assay

≥98% (HPLC)

form

powder

mol wt

536.87

storage condition

protect from light

solubility

THF: 1 mg/mL
chloroform: 5 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

shipped in

dry ice

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C\C=C\C(C)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(C)\C=C\C=C(/C)CC\C=C(\C)C

InChI

1S/C40H56/c1-33(2)19-13-23-37(7)27-17-31-39(9)29-15-25-35(5)21-11-12-22-36(6)26-16-30-40(10)32-18-28-38(8)24-14-20-34(3)4/h11-12,15-22,25-32H,13-14,23-24H2,1-10H3/b12-11+,25-15+,26-16+,31-17+,32-18+,35-21+,36-22+,37-27+,38-28+,39-29+,40-30+

InChI key

OAIJSZIZWZSQBC-GYZMGTAESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lycopene, an acyclic carotenoid pigment, is a bioactive natural compound commonly produced by many red fruits and vegetables such as watermelon, pink guava, and tomato, and photosynthetic algae. Current research suggests that this plant metabolite acts as an inhibitor and may exhibit diverse biological activities, including antioxidant, anti-carcinogenic, radioprotective, anticancer and anti-inflammatory properties.

Application

Lycopene has been used:
  • to analyze its chemo-preventive effects on cutaneous tumors in in vitro mouse epidermal cell line[1]
  • as a reference standard for quantitative high-performance liquid chromatography (HPLC) to analyze antioxidant capacity of endogenous carotenoids in mammalian cell lines[2]
  • as a reference standard in high-performance liquid chromatography (HPLC) to identify carotenoids from engineered Saccharomyces cerevisiae[3]

Lycopene is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and drug discovery research.

Biochem/physiol Actions

Antioxidant micronutrient of tomatoes associated with decreased risk for cancer and cardiovascular disease. Enhances gap juction communication between cells via upregulation of connexin 43 and reduces proliferation of cancer cells in culture. Inhibits cholesterol synthesis and enhances low-density lipoprotein degradation.
Lycopene has been found to be active against oral cancer[4] and to inhibit metastasis of human liver adenocarcinoma cells by down regulation of NOX4.[5] Lycopene was also found to be a beneficial supplement for people with cardiovascular diseases.[6][7]

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonia Gupta et al.
International journal of preventive medicine, 6, 70-70 (2015-09-04)
Without pigments, we are nothing. Life presents us with a kaleidoscope of colors. From the green grass of home to a forest's ruddy autumn hues, we are surrounded by living colors. Living things obtain their colors, with few exceptions, from
Bo-Yi Jhou et al.
Journal of agricultural and food chemistry, 65(32), 6893-6903 (2017-07-21)
NADPH oxidase 4 (NOX4), with the sole function to produce reactive oxygen species (ROS), can be a molecular target for disrupting cancer metastasis. Several studies have indicated that lycopene exhibited anti-metastatic actions in vitro and in vivo. However, the role
High-level beta-carotene production from xylose by engineered Saccharomyces cerevisiae without overexpression of a truncated HMG1 (tHMG1)
Sun L, et al.,
Biotechnology and Bioengineering, 3522-3532 (2020)
Subramaniam Pennathur et al.
Free radical biology & medicine, 49(2), 205-213 (2010-04-15)
Lycopene, a carotenoid found in tomatoes, is a proven antioxidant that may lower the risk of certain disorders including heart disease and cancer. Hypochlorous acid (HOCl) is an oxidant linked to tissue oxidation in cardiovascular disease and other inflammatory disorders
A V Rao et al.
Journal of the American College of Nutrition, 19(5), 563-569 (2000-10-07)
Lycopene, a carotenoid without provitamin-A activity, is present in many fruits and vegetables; however, tomatoes and processed tomato products constitute the major source of lycopene in North American diet. Among the carotenoids, lycopene is a major component found in the

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How can this product be dissolved? Is this dissolved in water, ethanol, methanol, isopropanol, acetonitrile, or others?

    1 answer
    1. The solubility of this product has been determined in THF: 1 mg/mL and chloroform: 5 mg/mL. Other solvents have not been tested.
      The Merck Index indicates the following solvents:
      carbon disulfide (1g/50ml)
      boiling ether (1g/3L)
      boiling petroleum ether (1g/12 L)
      hexane at 0 °C (1g/14L)
      almost insoluble in methanol or ethanol
      soluble in chloroform or benzene.

      The solubility in DMSO has been referenced in the publications below:
      10 mM in DMSO
      Am J Physiol Endocrinol Metab 293:E1820-E1827, 2007
      http://ajpendo.physiology.org/content/293/6/E1820.full
      100 mM in 10% DMSO/PBS
      Iranian Journal of Pharmaceutical Research (2007), 6 (3): 173-177
      http://www.ijpr-online.com/docs/20073/IJPR425.pdf
      solubilized in DMSO
      Mutation Research/Genetic Toxicology and Environmental Mutagenesis, 416(1-2): 125-128, 1998

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service