Skip to Content
Merck
All Photos(1)

Key Documents

SMB00570

Sigma-Aldrich

Methyl asterrate

≥95% (LC/MS-UV)

Synonym(s):

methyl 2-hydroxy-6-(4-hydroxy-2-methoxy-6-methoxycarbonylphenoxy)-4-methylbenzoate

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$496.00

$496.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$496.00

About This Item

Empirical Formula (Hill Notation):
C18H18O8
CAS Number:
Molecular Weight:
362.33
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

$496.00


Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥95% (LC/MS-UV)

form

solid

solubility

DMSO: 1 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC1=CC(OC)=C(OC2=C(C(O)=CC(C)=C2)C(OC)=O)C(C(OC)=O)=C1

InChI

1S/C18H18O8/c1-9-5-12(20)15(18(22)25-4)13(6-9)26-16-11(17(21)24-3)7-10(19)8-14(16)23-2/h5-8,19-20H,1-4H3

InChI key

KYDNOVLBVBYOSW-UHFFFAOYSA-N

General description

Natural product derived from fungal source.

Biochem/physiol Actions

GPCR ligand

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiang-Yun Zhang et al.
Chinese journal of natural medicines, 10(1), 36-39 (2013-01-11)
To investigate the chemical constituents of Allophylus longipes. Compounds were isolated and purified by various chromatographic techniques and their structures were elucidated by physicochemical characteristics and spectral data. Twenty-five compounds were isolated and identified as cycloart-24-en-3β, 26-diol (1), 3-oxotrirucalla-7, 24-dien-21-oic
M J Fang et al.
Natural product research, 26(13), 1224-1228 (2011-11-03)
A new diphenyl ether methyl 2-(2-formyl-3-hydroxy-5-methylphenoxy)-5-hydroxy-3-methoxybenzoate (3), together with four known compounds, asterric acid (1), methyl asterrate (2), 9(Z),12(Z)-nonadecadienoic acid (4) and orsellinic acid (5), were isolated from the Phoma sp. strain SHZK-2, which was isolated from a polluted environment

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service