Skip to Content
Merck
All Photos(1)

Key Documents

SMB00501

Sigma-Aldrich

Ethyl Tricosanoate

≥99% (GC)

Synonym(s):

Tricosanoic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H50O2
CAS Number:
Molecular Weight:
382.66
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥99% (GC)

form

powder

functional group

ester

lipid type

saturated FAs

shipped in

ambient

storage temp.

room temp

SMILES string

CCOC(CCCCCCCCCCCCCCCCCCCCCC)=O

InChI

1S/C25H50O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25(26)27-4-2/h3-24H2,1-2H3

InChI key

URGDXLFVAIKCMI-UHFFFAOYSA-N

General description

Ethyl Tricosanoate is an ethyl ester of tricosanoic acid. It has been reported to be one of the ester components responsible for the flavor of scotch whisky.

Application

Ethyl Tricosanoate may be employed as an internal standard in the following studies:
  • Quantification of fatty acids in squid visceral oil ethyl esters (SVOEE) isolated from Illex argentinus and Ommastrephes bartrami by gas chromatography.
  • Modified Folch procedure for the extraction of plasma lipids.
  • Estimation of fatty acid ethyl esters (FAEEs) in biodiesel by gas chromatography.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert J Pawlosky et al.
Pediatric research, 60(3), 327-333 (2006-07-22)
Efficacy of (13)C-U-18:2n-6 and (2)H(5)-20:3n-6 toward synthesis of labeled-20:4n-6 was studied in newborn infants utilizing compartmental models of plasma labeled n-6 fatty acids (FA). Ten infants received oral doses of (13)C-U-18:2n-6 and (2)H(5)-20:3n-6 ethyl esters (100 and 2 mg/kg, respectively).
Fractionation of squid visceral oil ethyl esters by short-path distillation.
Liang JH and Hwang LS.
Journal of the American Oil Chemists' Society, 77(7), 773-777 (2000)
Fractionation of urea-pretreated squid visceral oil ethyl esters.
Hwang LS and Liang JH.
Journal of the American Oil Chemists' Society, 78(5), 473-476 (2001)
Charalambous G.
The Quality of foods and beverages V1: Chemistry and technology, 209-209 (2012)
Performance of whole-cells lipase derived from Mucor circinelloides as a catalyst in the ethanolysis of non-edible vegetable oils under batch and continuous run conditions.
Carvalho AKF, et al.
Industrial Crops and Products, 67, 287-294 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service