Skip to Content
Merck
All Photos(1)

Documents

SMB00446

Sigma-Aldrich

Peimine

≥98% (HPLC)

Synonym(s):

(3β,5α,6α)-Cevane-3,6,20-triol, Dihydroisoimperialine, Verticine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H45NO3
CAS Number:
Molecular Weight:
431.65
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

mp

223 °C

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

C[C@H]1CC[C@@]2([H])[C@](O)(C)[C@@]3([H])CC[C@@]4([H])[C@]5([H])C[C@H](O)[C@@]6([H])C[C@@H](O)CC[C@]6(C)[C@@]5([H])C[C@@]4([H])[C@]3([H])CN2C1

InChI

1S/C27H45NO3/c1-15-4-7-25-27(3,31)21-6-5-17-18(20(21)14-28(25)13-15)11-22-19(17)12-24(30)23-10-16(29)8-9-26(22,23)2/h15-25,29-31H,4-14H2,1-3H3/t15-,16-,17+,18+,19-,20-,21-,22-,23+,24-,25-,26+,27-/m0/s1

InChI key

IUKLSMSEHKDIIP-BZMYINFQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Peimine is an alkaloid of Traditional Chinese Medicine that produces anti-inflammatory properties.

Biochem/physiol Actions

Peimine is a known MAPK inhibitor, as well as an inhibitor of p38, ERK and JNK phosphorylation, and NFκB activation. Peimine shows antitussive, expectorant and anti-inflammatory effects in vivo.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service